CTS™ L-Glutamine (200 mM) (100X)
Gibco™

CTS™ L-Glutamine (200 mM) (100X)

Gibco CTS L‐Glutamine is a ready-to-use 200 mM stock solution that acts as a cell culture supplement. L-glutamine is an amino acid that is required for cell culture.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A4000225302100 mLBag - BioProcess Container (BPC)
A4000225301100 mLBottle
A4000225303200 mLBag - BioProcess Container (BPC)
Catalog number A4000225302
Price (CLP)
-
Quantity:
100 mL
Format:
Bag - BioProcess Container (BPC)

Gibco Cell Therapy Systems (CTS) L‐Glutamine is a ready-to-use 200-mM stock solution that acts as a cell culture supplement. L-glutamine is an amino acid that is required for cell culture. It is involved in the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. Unlike most other amino acids, L-glutamine is not stable in solution. The rate at which degradation proceeds is a function of time, temperature, and pH.

Closed systems compatible
CTS L-Glutamine is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable PVC/C-Flex hybrid tubing with a female Luer lock that helps provide versatile connection options for integrating into a closed system manufacturing process.

CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Concentration200 mM (100X)
Culture TypeMammalian and Stem Cell Culture
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBag - BioProcess Container (BPC)
Manufacturing QualityISO13485, MDSAP, FDA-registered, 21CFR820
Quantity100 mL
Shelf Life12 Months from Date of Manufacture
Shipping ConditionDry Ice
FormClear, colorless solution
Product TypeCell Culture Supplement
SterilitySterile-filtered
Sterilization Method0.2 μm Sterile-filtered
Unit SizeEach
Contents & Storage
-20°C to -5°C. Protect from light.