Product Image
Applied Biosystems™
MicroSEQ™ ID Fungal ITS Gene Library v1.0
The MicroSEQ™ ID ITS Fungal Gene Library v1.0 is a curated collection of Internal Transcribed Spacer (ITS) sequences designed for accurate identification of fungal organisms. This library targets the highly polymorphic ITS region to enable high resolution species level identification of closely related or rare environmental fungi.
Have Questions?
Catalog NumberQuantity
A400054331 Ea.
Catalog number A40005433
Price (CLP)
-
Quantity:
1 Ea.
The MicroSEQ™ ID ITS Fungal Gene Library v1.0 is a curated collection of Internal Transcribed Spacer (ITS) sequences designed for accurate identification of fungal organisms. This library targets the highly polymorphic ITS region to enable high resolution species level identification of closely related or rare environmental fungi.
Summary:
  • Target Region: Internal Transcribed Spacer (ITS)
  • Library Size: 38,229 sequence entries
  • Taxonomic Breadth: 3,686 genera and 22,265 fungal species
  • Compatibility: Designed for use with MicroSEQ™ ID Analysis Software
Features
Comprehensive Coverage
Includes over 38,000 validated sequence entries spanning more than 22,000 fungal species, significantly reducing the likelihood of 'no-match' results for environmental and rare isolates.
High Discriminatory Power
The ITS region is highly polymorphic, enabling precise differentiation at the species level where conserved regions like the D2 (LSU) may lack sufficient resolution.
Curated and Validated Content
Sequence entries are meticulously curated to enable high standards of accuracy and consistency, offering reliable fungal identification for regulated environments.
Enhanced Identification Capability
Specifically designed to support the identification of closely related, rare, and environmental fungal isolates often encountered in industrial and environmental monitoring.
For Research Use Only.
Specifications
Product LineMicroSEQ
Product TypeLibrary
Quantity1 Ea.
Shipping ConditionRoom Temperature
SoftwareMicroSEQ™ ID Analysis Software
Target Organism ClassFungi
TechniqueSBT (Sequence-Based Typing)
FormatKit
Isolation TechnologyTube Centrifugation
Sequencing TypeDNA and RNA Sequencing
Unit SizeEach