Now in Stock: Gibco Anti-Anti (100X). Order now >
Product Image
Gibco™

Anti-Anti Replacement Kit

This product is being provided as an alternative to Antibiotic-Antimycotic (100X) (Cat. No. 15240062) as we continue to build inventory.
Have Questions?
Catalog NumberSterility
A40006088Sterile-filtered
Catalog number A40006088
Price (HKD)
842.00
Each
Sterility:
Sterile-filtered
Price (HKD)
842.00
Each

This Antibiotic-Antimycotic Replacement Kit is offered as an equivalent product to Antibiotic-Antimycotic (100X) (Cat. No. 15240062). It contains two bottles of Penicillin-Streptomycin (10,000 U/mL) (100 mL) (Cat. No. 15140122) and one bottle of Amphotericin B (20 mL) (Cat. No. 15290018).

This kit can be used to supplement the same volume of medium as 200 mL of Antibiotic-Antimycotic (100X). To achieve equivalent performance as Antibiotic-Antimycotic (100X), follow the 'Anti-Anti Replacement Protocol' found below.

For Research Use Only.
Specifications
Concentration100X, 1000X - See 'Anti-Anti Replacement Protocol' for details.
Shelf Life12 Months
FormClear, colorless to slight yellow solution
Product TypeAntibiotic-Antimycotic
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesNone
Without AdditivesNone
Unit SizeEach