Pharmaceutical Analytics QuantStudio™ 7 Pro Real-Time PCR System, 96-well, 0.1 mL
Applied Biosystems™

Pharmaceutical Analytics QuantStudio™ 7 Pro Real-Time PCR System, 96-well, 0.1 mL

The Pharmaceutical Analytics QuantStudio 7 Pro Real-Time PCR System is designed for users who need superior performance and security options in an innovative real-time PCR system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExternal Computer
A40006420Laptop
A40006425Not Included
A40006426Desktop
Catalog number A40006420
Price (TWD)
-
External Computer:
Laptop

The Applied Biosystems Pharmaceutical Analytics QuantStudio 7 Pro Real-Time PCR System is used together with specialized sample preparation and quantitation kits for host cell protein, host cell DNA, and Protein A impurity testing of biological products produced in cell culture.

The system adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. With facial authentication, Smart Help, multiplex capability, motorized interchangeable blocks, intuitive software, automation capabilities, and large capacitive touch-screen interface, the QuantStudio 7 Pro system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation. Designed for ultimate ease-of-use, the system offers high quality, high throughput, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.

Features of the Pharmaceutical Analytics QuantStudio 7 Pro Real-Time PCR System include:

  • Validated for use with AccuSEQ Real-Time PCR Detection Software v4.0
  • Integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, resDNASEQ, and ProteinSEQ assays
  • Built-in software features to support compliance with 21 CFR Part 11 guidelines
  • Peace of mind with Smart Help and Smart Remote Support; contact Technical Support directly from the touchscreen
  • Application versatility with easily changed blocks and four different block types
  • Increased productivity with automation compatibility for 24/7 operation
  • Remote monitoring of run status using the Instrument Connect monitoring app
  • Design and Analysis software compatible with PC and Mac operating systems

Pharmaceutical analytics real-time PCR applications

When used with AccuSEQ Real-Time PCR Detection Software and SEQ analytical testing assays, the Pharmaceutical Analytics QuantStudio 7 Pro system employs rapid and highly sensitive molecular methods to efficiently detect impurities, identify contaminants, and quantify titer across various applications.

Peace of mind with Smart service

Minimize downtime with premium service offerings for the QuantStudio 7 Pro system. Smart Help allows users to contact Technical Support directly from the touchscreen, without the need to download files or spend time on the phone. Experienced technicians will analyze the request and respond according to the user's preference (phone or email) and resolve the issue as quickly as possible.

Smart Remote Support is an immersive collaboration tool that allows customers to quickly and efficiently show issues to Technical Support for resolution, helping reduce instrument downtime compared to on-site repair and helping minimize the need for on-site visits.

Connect

Our cloud-based Connect platform provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the platform are web browser-based software tools that are flexible, fast, and easy to use. These software tools support a variety of analyses, including relative quantitation, standard curve, high resolution melt (HRM), genotyping, and presence/absence. They are compatible with Mac or PC, as well as most Applied Biosystems instrument file types.

Format flexibility

The QuantStudio 7 Pro system accommodates the interchange of 96-well (0.2 mL), 96-well (0.1 mL), 384-well, and TaqMan array card (TAC) blocks. The system is delivered with one block type; other block types may be purchased separately. In addition, an Orbitor Microplate Mover is available for this system to support high-throughput workflows.

Accomplish more, faster

Run hundreds of real-time PCR reactions effortlessly using TaqMan Array 384-well microfluidic cards. And when integrated with the Orbitor Microplate Mover (available separately), the QuantStudio 7 Pro system maximizes your throughput capabilities.

High application versatility

Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, and gene regulation.

Results you can trust

Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex system, which features six decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.

Sensitive data analysis

Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
CommunicationsThermo Fisher Connect Platform, USB, LAN, or WiFi
Dimensions54.7 x 33.8 x 52.5 cm (H x W x D)
Display Type12 in. Color Touchscreen
External ComputerLaptop
Quantity1 System
Shipping ConditionRoom Temperature
Voltage100/240 V ±10%
Weight38 kg
Block Configurations96-well, 6-Zone VeriFlex Block
Block FormatInterchangeable
Capacity96-well, 0.1 mL
Product TypeqPCR System
Unit SizeEach