POROS™ GoPure™ AAV9 Column
Thermo Scientific™

POROS™ GoPure™ AAV9 Column

POROS GoPure AAV9 chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAV9 columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDiámetro interior (métrico)Longitud (métrico)Volumen del lecho
A4001173214 cm10 cm1540 mL
A658722.5 cm5 cm25 mL
A658732.5 cm10 cm50 mL
A658805 cm5 cm100 mL
A658815 cm10 cm200 mL
A658828 cm5 cm250 mL
A658838 cm10 cm500 mL
A6588410 cm5 cm390 mL
A6588510 cm10 cm785 mL
A4001173114 cm5 cm770 mL
A4001173720 cm5 cm1570 mL
A4001173420 cm10 cm3140 mL
Número de catálogo A40011732
Precio (USD)
-
Diámetro interior (métrico):
14 cm
Longitud (métrico):
10 cm
Volumen del lecho:
1540 mL

POROS GoPure AAV9 chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAV9 columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards. Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ CaptureSelect™ AAV9 affinity resin, POROS GoPure AAV9 columns offer high specificity and capacity for AAV9 viral vectors over a wide range of flow rates.

  • Pressure rating (bar/psi): 4/58
  • Operating temperature: 2–30°C
  • Bed support size: 12 μm
  • Inlet/outlet internal diameter: 2.00 mm (2.5 cm), 3.45 mm (5 cm), 4.57 mm (8 cm), 6.35 mm (10 cm, 14 cm, 20 cm)
  • Inlet/outlet port connections: 3/4 inch mini tri-clamps
  • Includes a 30 mL resin side sample for incoming QC

Features of POROS CaptureSelect AAV9 resin include one-step AAV9 purification from crude material with high purity, with high specificity and capacity, improving yield by reducing process steps. Free of animal-origin components, the CaptureSelect AAV9 ligand is a 12 to 15 kDa single-domain antibody fragment produced in the yeast Saccharomyces cerevisiae.

For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to human or animals.
Especificaciones
Volumen del lecho1540 mL
Cantidad1 Column
Diámetro interior (métrico)14 cm
Longitud (métrico)10 cm
TipoChromatography Column
Unit SizeEach