AmpSure™ Universal Probe qPCR Master Mix
Thermo Scientific™

AmpSure™ Universal Probe qPCR Master Mix

Thermo Scientific™ AmpSure™ Universal Probe qPCR Master Mix is a ready-to-use, 2X concentrated master mix for probe-based qPCR detection and quantitation of a variety of DNA targets. Designed to support gene expression, copy number variation (CNV), single nucleotide polymorphism (SNP) genotyping, and microbial and pathogen detection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A400130161 mL
A400130175 mL
A4001301510 mL
Catalog number A40013016
Price (EUR)
55,10
Each
Quantity:
1 mL
Price (EUR)
55,10
Each

Thermo Scientific™ AmpSure™ Universal Probe qPCR Master Mix is a ready-to-use, 2X concentrated master mix for probe-based qPCR detection and quantitation of a variety of DNA targets. Designed to support gene expression, copy number variation (CNV), single nucleotide polymorphism (SNP) genotyping, and microbial and pathogen detection. AmpSure Universal Probe qPCR Master Mix is a versatile master mix that can support multiple qPCR applications, instrument platforms, and suppliers' assays; simplifying reagent selection and laboratory workflows.

Features:

  • Competitive price per reaction for efficient qPCR workflows¹
  • Reliable multiplex gene expression analysis under fast and standard cycling conditions, helping to deliver PCR efficiencies up to 98% and R² values ≥0.999.
  • Precise SNP genotyping across fast and standard cycling protocols, designed to support up to 100% call rate and concordance results.
  • Accurate copy number variation analysis across multiple gene targets, helping to deliver up to 100% call rate.
  • Reliable analytical detection of pathogens and microbial DNA, designed to support ∣ΔCt∣ values as low as 0.01 and R² values up to 0.98.²
  • Performs on a wide range of leading qPCR instrument platforms
  • Compatible with TaqMan™ qPCR Assays and assays from other suppliers
  • Contains ROX™ passive reference dye for normalization on compatible instruments and heat-labile uracil-DNA glycosylase (UNG) to help reduce carryover contamination

Reliable Gene Expression Analysis

  • AmpSure Universal Probe qPCR Master Mix demonstrates sensitive and reliable gene expression analysis verified on several qPCR instrument platforms in fast and standard cycling protocols. The formulation is optimized for efficient amplification of multiplex (duplex) gene expression targets.

Precise SNP genotyping

  • AmpSure Universal Probe qPCR Master Mix demonstrates effective allelic discrimination in both fast and standard cycling conditions with both DME and non-DME assays. Using purified genomic DNA, the product helps deliver high call rate and accurate genotyping on a wide range of qPCR instrument platforms.

High accuracy CNV analysis

  • AmpSure Universal Probe qPCR Master Mix enables reliable copy number determination across multiple gene targets with accurate quantification in samples containing one to three copies. The product demonstrates consistent and accurate CNV analysis across a variety of qPCR instrument platforms.

Supports microbial and pathogen analytical detection

  • AmpSure Universal Probe qPCR Master Mix supports the analytical detection of microbial and pathogen DNA, with amplification of one or two targets across multiple qPCR platforms. Multiplex performance may require assay-specific optimization.

Compatible with multiple suppliers' assays

  • AmpSure Universal Probe qPCR Master Mix is compatible with a range of singleplex and duplex assay designs and chemistries. The universal formulation produces reliable results with both TaqMan and IDT™ assays when compared to IDT PrimeTime™ Gene Expression Master Mix in identical workflow configurations.

The master mix uses TaqMan fluorogenic 5' nuclease chemistry. During amplification, a target-specific probe containing reporter and quencher dyes anneals to the target sequence and is cleaved by the 5' nuclease activity of Taq DNA polymerase. Cleavage separates the reporter from the quencher, increasing fluorescence as amplification proceeds; the resulting fluorescence increase is proportional to the amount of amplicon produced.

AmpSure Universal Probe qPCR Master Mix is a versatile master mix that can help simplify protocol development and workflow integration, while reducing the potential for unused or expired application-specific reagents and incorrect master mix selection. Details of each of the qPCR application workflows, including recommended materials, sample preparation, reaction setup, and data analysis, are provided in the AmpSure Universal Probe qPCR Master Mix user guide included.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions100 Reactions
Product TypeqPCR Master Mix
Quantity1 mL
Concentration2X (Master Mix)
Detection MethodqPCR
For Use With (Application)Real Time PCR (qPCR), Gene Expression, Genotyping, Copy Number Variation Analysis, Pathogen Detection
FormLiquid
GC-Rich PCR PerformanceMedium
PCR Method2-step RT-qPCR, qPCR, Multiplex qPCR
Reaction SpeedFast or Standard
Unit SizeEach