How do I set up methods on my Thermal Cycler Fleet Control Software?
Within the Thermal Cycler Fleet Control Software, go to Settings, then Help Tutorials, where you can find tutorials on setting up runs and methods.
How do I connect my Thermal Cycler Fleet Control Software to the network?
Within the Thermal Cycler Fleet Control Software, go to Settings, then Help Tutorials, where you can find tutorials on network configuration.
How do I create and manage users in the Thermal Cycler Fleet Control Software?
Within the Thermal Cycler Fleet Control Software, go to Settings, then Help Tutorials, where you can find tutorials on creating users and assigning user permissions.
Where is the manual for the Thermal Cycler Fleet Control Software?
The Quick Resource Card for the Fleet Control Software can be found here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/100077923_ThermalCyclerFleetSftwre_QR.pdf.
How can I monitor my thermal cycler remotely without being on the Cloud?
The Thermal Cycler Fleet Control Software allows monitoring of the following instruments from within a firewall:
SimpliAmp Thermal Cycler
ProFlex PCR System
Veriti Thermal Cycler
VeritiPro Thermal Cycler
Automated Thermal Cycler (ATC)
Which thermocyclers can be controlled using the Thermal Cycler Fleet Control Software?
The Thermal Cycler Fleet Control Software is compatible with the following instruments:
- SimpliAmp Thermal Cycler
- ProFlex PCR System
- Veriti Thermal Cycler
- VeritiPro Thermal Cycler
- Automated Thermal Cycler (ATC)
- MiniAmp Thermal Cycler
- MiniAmp Plus Thermal Cycler