Genexus™ Barcodes 1-96 AS - FAQs
Genexus™ Barcodes 1-96 AS - FAQs
View additional product information for Genexus™ Barcodes 1-96 AS - FAQs (A40257)
31 product FAQs found
What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?
Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.
Which extraction kits are compatible with Oncomine Myeloid Research Assay GX?
Here is a list of kits that are compatible with Oncomine Myeloid Research Assay GX:
Nucleic acid isolation - RNA samples
- MagMAX mirVana Total RNA Isolation Kit, Cat. No. A27828
- PureLink Total RNA Blood Kit, Cat. No. K156001
- RNaseZap RNase Decontamination Solution, Cat. No. AM9780, AM9782, AM9784
Nucleic acid isolation - DNA samples
- MagMAX DNA Multi-Sample Ultra 2.0 Kit, Cat. No. A36570
- PureLink Genomic DNA Mini Kit, Cat. No. K1820-01
What are the sample input requirements for Oncomine Myeloid Research Assay GX?
The recommended minimum sample input is 27.75 ng of DNA (1.11 ng/µL minimum concentration) and 14.25 ng of RNA (0.95 ng/µL minimum concentration).
What sample controls should I use with the Oncomine Myeloid Research Assay GX?
The recommended controls are:
DNA Controls:
- Seraseq® Myeloid Mutation DNA Mix, Cat. No. 0710-0408 (SeraCare®)
- Myeloid DNA Reference Standard, Cat. No. HD829 (Horizon™)
- AcroMetrix™ Oncology Hotspot Control, Cat. No. 969056
RNA Controls:
- Universal Human Reference RNA, Cat. No. 740000 (Agilent™)
- Seraseq® Myeloid Fusion RNA Mix, Cat. No. 0710-0407 (SeraCare®)
How many Oncomine Myeloid Research Assay GX samples can be multiplexed on a GX5 Chip?
Multiplex sequencing of up to 8 Oncomine Myeloid Research Assay GX samples (8 DNA+ 8 RNA) per lane on a GX5 Chip can be performed in a single run. There are 4 lanes on the GX5 Chip, so 32 total samples (32 DNA+ 32 RNA) can be run on one GX5 Chip (8 samples per run).
What are the components of Oncomine Myeloid Research Assay GX?
The Oncomine Myeloid Research Assay GX (Cat. No. A47857) includes the 2-pool DNA panel, the 1-pool RNA panel, Genexus Strip 1, and Genexus Strip 2-AS. The DNA panel is provided in two packs of 8 tubes (4 tubes of Myeloid DNA Pool 1 and 4 tubes of Myeloid DNA Pool 2 per pack). The RNA panel is provided in one pack of 8 tubes (Myeloid RNA Pool 1). Each primer pool in the panel is provided in pairs of tubes, where each tube pair contains one tube with primers in position 1 and one empty uncapped tube in position 2. Three 8-strip packs of the Genexus Strip 1 and Genexus Strip 2-AS (24 total of each strip) are provided with each kit.
How many samples can I run using Oncomine Myeloid Research Assay GX?
The contents of each Oncomine Myeloid Research Assay GX kit are sufficient for up to 32 samples (32 x 3‑pool reactions: 32 x 2‑pool DNA reactions and 32 x 1‑pool RNA reactions).
How many primer pools are present in the Oncomine Myeloid Research Assay GX?
The Oncomine Myeloid Research Assay GX includes 3 pools of Ion AmpliSeq oligonucleotide primers (a 2-pool DNA panel and a 1-pool RNA panel).
What is the Oncomine Myeloid Research Assay GX gene content?
The Oncomine Myeloid Research Assay GX targets key genes and fusions associated with major myeloid disorders, including acute myeloid leukemia (AML), myeloid dysplastic syndrome (MDS), myeloproliferative neoplasms (MPN), chronic myeloid leukemia (CML), chronic myelomonocytic leukemia (CMML), and juvenile myelomonocytic leukemia (JMML). The DNA panel comprises 40 key genes while the RNA panel includes a broad fusion panel of 29 driver genes, enabling detection over 600 unique fusion isotypes. See flyer for gene targets (https://assets.thermofisher.com/TFS-Assets/CSD/Flyers/Oncomine_myeloid_assay_gx_flyer.pdf).
What is the Oncomine Myeloid Research Assay GX designed for ?
The Oncomine Myeloid Research Assay GX is a comprehensive targeted next-generation sequencing (NGS) assay designed for sensitive detection of myeloid disorder-associated DNA mutations and fusion transcripts in blood and bone marrow samples.
How many samples of Oncomine Precision Assay GX can I run on each lane of the GX5 Chip?
The capacity of the GX5 Chip varies depending on the sample type:
- For plasma cfTNA samples, the workflow is configured to accommodate 1 sample per GX5 Chip lane (or 4 samples on a single GX5 Chip).
- For FFPE DNA+RNA samples, the workflow is configured to accommodate 4 samples per GX5 Chip lane.
- For FFPE DNA-only or RNA-only samples, the workflow is configured to accommodate 8 samples per GX5 Chip lane.
How many sequencing reactions can I perform with Oncomine Precision Assay GX (Cat. No. A44350)?
The maximum number of samples that can be analyzed per kit depends upon the type of analysis that you want to perform. Each kit provides the targeted pancancer panel and library reagents sufficient to perform up to 32 sequencing reactions on the Genexus Integrated Sequencer.
- Simultaneous DNA- and RNA-based variant analysis for up to 16 FFPE tissue samples (one DNA reaction and one RNA reaction per sample)
- DNA-only or RNA-only variant analysis for up to 32 FFPE tissue samples
- Cell-free TNA (cfTNA) variant analysis (combined DNA and RNA) for up to 32 plasma samples
How much of each sample do I need for the Oncomine Precision Assay GX?
For solid tumor samples, the optimal input amount is 10 ng with validated input in the range of 5-15 ng. For liquid biopsy samples, the optimal input amount is 20 ng with validated input in the range of 5-40 ng.
For optimal results, load 20 µL of input with a concentration of ≥0.67 ng/µL for FFPE DNA and FFPE RNA samples and 20 µL of input with a concentration of ≥1.33 ng/µL for cfTNA samples. If the concentration of the sample is lower than optimal, use as much input as possible for each reaction.
What controls should I use with Oncomine Precision Assay GX?
Here are the various controls we recommend depending upon the starting material:
FFPE DNA samples
- Structural Multiplex Reference Standard (FFPE), Cat. No. HD789 (Horizon™)
- EGFR Gene-Specific Multiplex Reference Standard (FFPE) 5% AF, Cat. No. HD300 (Horizon™)
- KRAS Gene-Specific Multiplex Reference Standard (FFPE), Cat. No. HD301 (Horizon™)
- Seraseq® Lung & Brain CNV Mix, + 3 copies, Cat. No. 0710-0414 (SeraCare®)
- Seraseq® Breast CNV Mix, + 6 copies, Cat. No. 0710-0412 (SeraCare®)
FFPE RNA samples
- Seraseq® FFPE Tumor Fusion RNA v4 Reference Material, Cat. No. 0710-0496 (SeraCare®)
- Seraseq® FFPE NTRK Fusion RNA Reference Material, Cat. No. 0710-1031 (SeraCare®)
cfTNA samples
- Multiplex I cfDNA Reference Standard Set, Cat. No. HD780 (Horizon™)
Which sample types can I use with Oncomine Precision Assay GX?
Oncomine Precision Assay GX is compatible with solid tumor samples (FFPE DNA or FFPE RNA) as well as liquid biopsy samples (cfTNA).
Which regions are targeted with Oncomine Precision Assay GX?
Oncomine Precision Assay GX is designed for the detection of biomarkers in genes such as EGFR, BRAF, KRAS, ALK, ROS1, NTRK, RET, HER2. The assay analyzes 78 variants, including mutations (45), CNVs (14), and fusion variants (19), across 50 key genes. Included are tumor suppressor genes such as TP53, cancer drivers, and resistance mutations. Content has been carefully curated to include all relevant targets and targets of emerging importance in precision oncology clinical research.
What is included in Oncomine Precision Assay GX (Cat. No. A46291)?
Oncomine Precision Assay GX (Cat. No. A46291) includes the following:
- OPA Pool 1 panel (Part No. A44350)
- Genexus Strip 1, quantity of 8 strips (Part No. A46812)
- Genexus Strip 2-HD, quantity of 8 strips (Part No. A46814)
- The OPA Pool 1 is provided in 8 pairs of tubes, where each tube pair contains one tube with forward (FWD) primers and one tube with reverse (REV) primers.
What is contained in Genexus Primer Pool Tubes (Cat. No. A40262)?
The Genexus Primer Pool Tubes kit contains 50 empty assemblies (2 tubes/assembly) of the custom tubes required to deliver primer oligos into the automated library preparation reaction. Each assembly is serialized for traceability and can be used with Ion AmpliSeq- and Ion AmpliSeq HD-based assays. Pipet an aliquot of panel primers from a custom panel or Ion AmpliSeq On-Demand Panel to run the assay on the Genexus Integrated Sequencer. The kit also contains a bag of 100 caps.
Note:
- Assays using Ion AmpliSeq chemistry have primer pools that are loaded in capped tubes in position 1 of the primer pool tube carriers. Empty uncapped tubes are loaded in position 2.
- Assays using Ion AmpliSeq HD chemistry have FWD and REV primer pools that are loaded in capped tubes in both positions of the primer pool tube carrier.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the storage conditions for Genexus Controls kit (Cat. No. A40267)?
We recommend storing Genexus Controls at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is contained in the Genexus Controls kit (Cat. No. A40267)?
The Genexus Controls kit provides library and template preparation controls for use with the Genexus Integrated Sequencer. Each kit contains an Ion AmpliSeq-based Genexus Control Panel-AS and a Genexus DNA Control, which can be used to prepare up to 32 automated control libraries. The kit also provides Genexus Control Library-AS, sufficient for four template preparations.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How should I prep Genexus Templating Strips 3 and 4-GX5 for use on the Genexus platform?
Remove the templating strips from their boxes in the refrigerator or freezer, and prepare them for loading in the sequencer.
- Genexus Strip 3-GX5 needs to equilibrate to room temperature for 30 mins.
- Genexus Strip 4 needs to thaw on ice for at least 30 mins. Keep the strips on ice until you load them in the sequencer.
Genexus Strip 3-GX5 contains magnetic beads in one or two positions, yellow or brown in color, that sometimes get trapped in the upper "keyhole" of the tube. Dislodge these beads from the keyhole before installing the strip in the sequencer. Use the following procedure for each strip:
- Invert the strip 3-4 times to dislodge beads that are trapped in the keyholes.
- To remove any remaining beads and liquid from the keyholes, grasp the strip at one end with the strip seal facing up, then swing the strip with a rapid, downward centrifugal arm motion, ending with a sharp wrist-flick.
- Grasp the strip at the other end, then repeat the centrifugal motion.
- Check tube positions for significant amounts of beads that are still trapped in keyholes, then repeat the centrifugal motion, if needed. It is acceptable if a few beads remain in the keyhole or on the tube wall, but most should be either in suspension or in a pellet at the bottom of the tube.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the storage conditions for Genexus Templating Strips 3 and 4-GX5 (Cat. No. A40263)?
Genexus Strips 3-GX5 (grey) should be stored at 4 degrees C and Genexus Strip 4 (yellow) should be stored at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What chemistry can I use Genexus Templating Strips 3 and 4-GX5 with?
Genexus Templating Strips 3 and 4-GX5 (Cat. No. A40263) can be used with both Ion AmpliSeq library chemistry and Ion AmpliSeq HD library chemistry runs.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is contained in the Genexus Templating Strips 3 and 4-GX5 (Cat. No. A40263)?
Genexus Templating Strips 3 and 4-GX5 (Cat. No. A40263) provides eight pairs of strips, required to perform automated template preparation and chip loading of two Ion Torrent GX5 chips using the Genexus Integrated Sequencer. The Genexus sequencer uses 2 strips (Genexus Strip 3-GX5 and Genexus Strip 4) to support template preparation. One template strip pair is required for every lane that is used in a run.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is included in Genexus Barcodes 65-96 AS (Cat. No. A40260)?
Genexus Barcodes 65-96 AS provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes AS 65-96) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is included in Genexus Barcodes 33-64 AS (Cat. No. A40259)?
Genexus Barcodes 33-64 AS provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes AS 33-64) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is included in Genexus Barcodes 1-32 AS (Cat. No. A40258)?
Genexus Barcodes 1-32 AS provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes AS 1-32) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Will the Genexus Software notify me if the barcodes had been previously used?
When necessary, the software will alert the user during the run plan setup that not enough barcodes are available.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long can the barcodes remain on the Genexus platform?
Barcodes plates can remain on the instrument deck until all barcodes are used. Once you remove the barcode plate, you cannot put it back on to use again as the instrument has no way of tracking barcode usage.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is included in Genexus Barcodes 1-96 AS (Cat. No. A40257) ?
The Genexus Barcodes 1-96 AS are supplied in 3 individual plates containing 32 dual barcodes per plate. The barcodes are lyophilized. Each barcode plate can also be ordered individually: Genexus Barcodes 1-32 AS (Cat. No. A40258), Genexus Barcodes 33-64 AS (Cat. No. A40259), and Genexus Barcodes 65-96 AS (Cat. No. A40260).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many pipette tips do I get with Genexus Pipette Tips (Cat. No. A40266)?
12 racks of pipette tips are provided, containing 96 tips per rack. The pipette tips can be stored at room temperature.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.