Product Image
Applied Biosystems™

QuantStudio™ 1 Real-Time PCR System, 96-well, 0.2 mL, desktop

The Applied Biosystems QuantStudio 1 Real-Time PCR System makes quality and reliability the practical choice. This affordable instrument offers basic features ideal for those new to qPCR or those who have a limited budget.
Have Questions?
Número de catálogoCantidad
A404281 system
Número de catálogo A40428
Precio (USD)
-
Cantidad:
1 system

The Applied Biosystems QuantStudio 1 Real-Time PCR System makes quality and reliability the practical choice. This affordable instrument offers basic features ideal for those new to qPCR or those who have a limited budget. Despite its affordability, the QuantStudio 1 system offers the quality, reliability, and advanced user experience of our QuantStudio family of instruments.

Features of the QuantStudio 1 system include:

  • Touchscreen – a large 8.5-inch interactive touchscreen
  • Fixed block – one fixed 96-well, 0.2-mL isothermal block with an easy-to-open manual drawer
  • Multiplexing capability – offers the three most commonly used excitation and emission filters: FAM, VIC, and ROX
  • Protocols – pre-optimized protocol templates
  • Software – intuitive software that supports analysis via web browser or desktop computer
  • Dyes – three filters calibrated for FAM/SYBR Green/SYTO 9, VIC/HEX/TET/JOE, and JUN/ROX/Texas Red dyes
  • Sensitivity – detect differences as small as 1.5-fold quantities in singleplex reactions

Thermo Fisher Connect
Thermo Fisher Connect with cloud-enabled services provides scientists with a secure place to store, analyze, and share data. Upload data from the instrument or upload your own files from anywhere you have web access. Applied Biosystems qPCR Analysis Modules within Thermo Fisher Connect are web browser-based software tools that are flexible, fast, and easy to use.

Easy to use
The QuantStudio 1 Real-Time PCR System is compact in form, factory calibrated, and delivered in user-friendly packaging to enable fast and simple installation for immediate use. The interactive touchscreen provides a modern usability experience, and the simplified instrument software/pre-optimized protocol templates allow users of any experience level to quickly set up and start a run.

Design and analysis software
Option 1: QuantStudio Design and Analysis cloud software

  • Supports web browser-based system configuration with PC or MAC computer
  • Streamlined software for improved usability and response time
  • Enables secure access of your data when and where you want through Thermo Fisher Connect
  • No software to install, no licenses and no versions to update

Option 2: QuantStudio Design and Analysis desktop software

  • Supports traditional co-located computer system configuration on Windows™ 7 and Windows™ 10 OS
  • Streamlined software for improved usability and response time
  • No licenses to purchase

Results you can trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with OptiFlex technology, which features three coupled excitation and emission filter channels with white LED.

Sensitive data analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.

High application versatility
Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, and more.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Fondo (métrico)50 cm
Tipo de pantalla8.5 in. Touchscreen
Ordenador externoDesktop
Para utilizar con (equipo)QuantStudio™
Formato96-well Plate
Altura (métrico)40 cm
Línea de productosApplied Biosystems™ QuantStudio
Cantidad1 system
Intervalo de volumen de reacción10 to 100 μL
Condiciones de envíoAprobado para su envío a temperatura ambiente o en hielo húmedo
Peso<26 kg
Anchura (métrico)27 cm
Configuraciones de bloques96-well
Formato de bloqueNo intercambiable
Unit SizeEach

Preguntas frecuentes

What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio Design & Analysis Software (v1.x)?

If you are analyzing results from the QuantStudio 1 Real-Time PCR System, QuantStudio 3 Real-Time PCR System, or QuantStudio 5 Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Design & Analysis Software (v1.x) following the instructions below:
1. Open the data file in the QuantStudio Design & Analysis Software (v1.x).
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply.

With the QuantStudio 1 instrument, why am I only able to export my run files to USB, whereas the cloud and network options are not available?

It is possible that the instrument is not connected to the computer and is not linked with the Thermo Fisher Cloud. Please see page 38 of the QuantStudio 1 Real-Time PCR System User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017853_QS1%20UG.pdf) for instructions to link an instrument profile with the Thermo Fisher Cloud. Although default installation instructions recommend using an Ethernet cable, it can also be done wirelessly using a High Power USB WiFi Module, Cat. No. A26774 (https://www.thermofisher.com/order/catalog/product/A26774).

If you experience connection issues between the instrument and computer, it might be useful setting a static IP address on both the instrument and computer. On the computer, you can use online help, such as this one (https://kb.netgear.com/27476/How-do-I-set-a-static-IP-address-in-Windows) to set the static IP address. Here are the details for the settings:

- Computer IP: 10.0.0.2
- Instrument IP: 10.0.0.3
- Subnet: 255.255.255.0
- DNS1: 8.8.8.8
- DNS2: 8.8.4.4
- Default gateway: 10.0.0.1

On the instrument, the IP address can be found under Settings and About Instrument. If you see an IP address but are still having network issues, try to PING the IP Address in a command window of the computer:

- Select the windows start button and select the RUN command.
- Open the Run command and type in CMD.
- This will open the Command window.
- Type the word "PING", space, followed by the IP address and press enter.
- You should see 4 packets of data sent, 4 received, and 0 lost.

If you PING the IP Address successfully (4 sent, 4 received, 0 lost), but still cannot connect, it means that one of the needed ports is still not open. Please contact your IT department and review the instrument checklist.
If you PING the IP address unsuccessfully, please send us the serial number of the instrument so that we can contact your local engineer.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Where can I find the software and newest firmware for the QuantStudio 1 instrument?

In an effort to reduce environmental impact, we no longer supply the software on the CD. Please download it from here (https://www.thermofisher.com/uk/en/home/global/forms/life-science/quantstudio-software-download.html).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.