Opti-Plex™ Complexation Buffer
Gibco™

Opti-Plex™ Complexation Buffer

Opti-Plex Complexation Buffer is a cGMP-manufactured, chemically defined, animal origin-free (AOF), methionine-free, biotin-free, low-protein transfection complexation buffer based on Opti-MEM I Reduced Serum Medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A4096801100 mL
A4096803500 mL
Catalog number A4096801
Price (USD)
59.75
Each
Quantity:
100 mL
Price (USD)
59.75
Each

Opti-Plex Complexation Buffer is a cGMP-manufactured, chemically defined, animal origin-free (AOF), methionine-free, biotin-free, low-protein transfection complexation buffer based on Opti-MEM I Reduced Serum Medium. It is designed for complexation of ExpiFectamine transfection reagents for both standard transfection applications and transfections to generate methionine-labeled proteins.

Opti-Plex Complexation Buffer features include:

  • Contains recombinant insulin, recombinant transferrin, and GlutaMAX Supplement
  • Does not contain methionine, biotin, or phenol red
  • A part of the ExpiFectamine293 Met (–) Transfection Kit (Cat. No. A39249)
For Research Use Only.
Specifications
Chemical Name or MaterialComplexation Buffer
Recommended Storage100 mL Opti-Plex Complexation Buffer, store at 2–8°C (protect from light)
For Use With (Application)Protein Expression
Physical FormLiquid
Quantity100 mL
Unit SizeEach