How can I remove old dye sets from the 3730xl DNA Analyzer (Cat. No. A41046) software?
You can remove old dye sets by deleting the spectral plate records associated with the spectral run. In the spectral viewer, under the list of calibrations for the dye sets, please note the dates when the spectral plates were run. In the plate manager, do an Advanced Search. In the first column, titled "Condition", under the row "Type", select "=", and in the second column, titled "Value 1", select "Spectral Calibration", then click on the search button. Highlight and delete the processed plates associated with the spectral dates.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.