ProFlex™ 3x32-Well PCR System will be discontinued in May 2027. The recommended replacement is the PowerFlex™ Thermal Cycler, 3x32-Well.
ProFlex™ PCR System + Plastics Packages, 3 x 32-well
Applied Biosystems™

ProFlex™ PCR System + Plastics Packages, 3 x 32-well

The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems instruments with the flexible configuration and control features that adapt to your research needs.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripción
A41182Includes MicroAmp 8-Tube Strips with Attached Domed Caps, 0.2 mL (125 strips)
A41183Includes 5 x MicroAmp TriFlex 3 x 32-well PCR Reaction Plates (100 plates total)
Número de catálogo A41182
Precio (CLP)
-
Descripción:
Includes MicroAmp 8-Tube Strips with Attached Domed Caps, 0.2 mL (125 strips)
The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems instruments with the flexible configuration and control features that adapt to your research needs. This package includes the system plus plastics (either MicroAmp 8-Tube Strips with Attached Domed Caps or MicroAmp TriFlex PCR Reaction Plates and MicroAmp 32-Well Clear Adhesive Films).

Proflex PCR System
• Interchange 96-well 0.2-mL block with another block type if throughput needs change
• Access system remotely (and conveniently) via mobile app
• Program instrument in seconds with simple-to-use touchscreen interface
• Simulate old instrument with Thermal Simulation modes
• Optimize PCR quickly and efficiently with better-than-gradient VeriFlex blocks

MicroAmp 8-Tube Strips with Attached Domed Caps
MicroAmp 8-Tube Strips with Attached Domed Caps feature individually attached domed caps, dual large end-tabs, and graduated 20-μL markers on each tube to help prevent cross-contamination, pipetting errors, and sample identification errors.
• Strip of eight tubes with attached domed caps that open and close independently to prevent contamination and evaporation
• Angled caps that provide efficient tube strip positioning in racks and thermal cycler blocks
• Etched A-H letter labeling on individual tubes that is visible when caps are open or closed
• Dual end tabs for proper labeling and easy handling
• Graduated 20-μL measuring markers on each tube for visual volume verification

MicroAmp TriFlex 3 x 32-Well PCR Reaction Plates
MicroAmp TriFlex 3 x 32-Well PCR Reaction Plates have a unique design, with three identical polypropylene 32-well PCR plates connected by dual side-tabs for use with 32-well and standard 96-well thermal blocks. The plates offer the same thin-wall construction as all other MicroAmp plastics, for excellent heat transfer from thermal block to sample, and they are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.
• Designed for use with the ProFlex 3 x 32-well PCR System and all Applied Biosystems standard 96-well thermal cyclers
• Easy-to-tear dual side-tabs for simple separation into three individual 32-well plates
• Semi-skirted for easy handling and labeling
• Standard 250 μL working volume, with a 300 μL maximum well volume when used with an adhesive film
• Alphanumeric labeling on each 32-well plate for easy well identification
• Certified human DNA-, DNase-, and RNase-free

MicroAmp 32-Well Clear Adhesive Films
MicroAmp 32-Well Clear Adhesive Film is designed specifically for use with MicroAmp TriFlex 3 x 32-Well PCR Reaction Plates. The film provides an alternative to cap strips and seals 32-well samples securely to prevent PCR sample evaporation and well-to-well contamination.
• Same product design as MicroAmp Clear Adhesive Film for 96-well plates, but pre-cut to fit 32-well plates
• Easy to apply and remove—simply peel off the backing and apply firmly to the 32-well plate

For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
DescripciónIncludes MicroAmp 8-Tube Strips with Attached Domed Caps, 0.2 mL (125 strips)
Dimensiones56,5 x 33 x 27,2 cm (22 x 13 x 10,6 in)
Tipo de pantallaPantalla táctil (LCD TFT de 8,4 pulg.)
Compatibilidad de alto rendimientoProtocolos manuales
Velocidad de rampa máxima6 °C/s (bloque), 4,4 °C/s (muestra)
Cantidad1 package
Velocidad de reacciónRápida, estándar
Exactitud térmica±0,25 °C (de 35 °C a 99,9 °C)
Intervalo térmicoDe 0 °C a 100 °C
Uniformidad térmica< 0,5 °C (20 s después de alcanzar 95 °C)
Interfaz de usuarioUSB, Wi-Fi, nube, Ethernet
Tensión100 - 240 V
GarantíaGarantía limitada
Peso18,8 kg (41 lb.)
Configuraciones de bloques3 bloques VeriFlex de 2 zonas de 32 pocillos
Capacidad96 pocillos, 0,2 ml
Tipo de productoSistema PCR
Unit SizeEach
Contenido y almacenamiento
• Sistema de PCR ProFlex de 3 x 32 pocillos
• Un juego de tiras de ocho tubos MicroAmp con tapones abovedados acoplados, 0,2 ml (125 tiras)

Preguntas frecuentes

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.