Product Image
Applied Biosystems™
TaqMan™ Array Respiratory Tract Microbiota Comprehensive Card
TaqMan Array Respiratory Tract Microbiota Comprehensive Card is an efficient, easy-to-use TaqMan Array Card (48-assay format) for the characterization ofRead more
Have Questions?
Catalog NumberQuantity
A412381 Plate
Catalog number A41238
Price (BRL)
-
Quantity:
1 Plate
TaqMan Array Respiratory Tract Microbiota Comprehensive Card is an efficient, easy-to-use TaqMan Array Card (48-assay format) for the characterization of key respiratory tract microbial targets through real-time PCR. This card contains TaqMan Microbial Assays for 42 microbial targets, plus Universal Extraction Organism Control (B. atropheus), Universal DNA Spike-In Positive Control (Xeno), 18s control and human RNase P RPPH1 control. Each TaqMan Array Card can be used for running 7 samples and 1 control sample.
Key Feature of TaqMan Array Cards:
• Pre-formatted and inventoried—each card contains a select group of TaqMan Microbial Assays pre-configured in a micro fluidic array, ready to ship to you
• Convenient and easy to use—get the performance you’ve come to expect from TaqMan Microbial Assays without the need for expensive robotics
• Reproducible results—get consistent, reliable data across samples, studies, and labs—the same high data quality from card-to-card and lot-to-lot—even with different operators
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineTaqMan
Quantity1 Plate
Shipping ConditionApproved for shipment at ambient temperature
For Use With (Application)Pathogen Detection, Virus Detection
Unit SizeEach
Contents & Storage
Store refrigerated at 4°C.