Product Image
Applied Biosystems™

TaqMan™ Microbe Assay Type 2, Small

The assay is designed to one or more microbial sequences to maximize coverage of the target organismRead more
Have Questions?
Catalog NumberQuantity
A41332S (250 reactions/250 μL), inventoried
Catalog number A41332
Price (USD)
365.00
Each
Quantity:
S (250 reactions/250 μL), inventoried
The assay is designed to one or more microbial sequences to maximize coverage of the target organism
Specifications
FormatTube
Internal Probe ModificationFAM (5'), MGB (Minor Groove Binder) (3')
No. of Reactions250 reactions
Product LineTaqMan
Product TypeMicrobe Assay
QuantityS (250 reactions/250 μL), inventoried
Unit SizeEach
Contents & Storage
One tube of assay

Storage recommendation is -20°C.