Search
Search

| Catalog Number | Quantity |
|---|---|
| A41390 | 1 Kit |
The Gibco™ Hepatic Spheroid Kit is the first off-the-shelf kit for generation of hepatic spheroids in your own lab.
What you get:
Please note that components are shipped separately due to differences in storage temperatures.
Simplified protocol
Cryopreserved cells can be thawed and plated at your convenience, in as little sa 60 minutes. Try it on your own, or request a demo with an experienced scientist.
Stringent quality control
Our high standards help ensure only the best spheroid-forming lots become available, maximizing your chance of success. In addition, each lot is tested for:
Important! For best results, do not attempt hepatic spheroid formation in low attachment plates from other sources. Hepatocyte lots are qualified in Nunclon Sphera plates only and use of other plates can lead to variability in spheroid formation.
Flexible readouts
Analyze single spheroids, or pool several for detection in assays such as gene expression, immunostaining, viability, or CYP activity.
For Research Use Only. Not for use in diagnostic procedures.
• Spheroid-qualified hepatocytes (1 mL, liquid N2 vapor phase only)
• Hepatocyte thaw medium (45 mL, 2°C to 8°C)
• Plating supplement (Cocktail A + Dexamethasone) (18 mL, 2°C to 8°C)
• Maintenance supplement (Cocktail B + Dexamethasone) (20 mL, 2°C to 8°C)
• Fetal bovine serum (2 x 25 mL, -5°C to -30°C)
• Williams' E Medium, no phenol red (2 x 500 mL, 2°C to 8°C. Protect from light.)
• 8 Nunclon Sphera 96-well U-bottom plates