Applied Biosystems™ RapidLINK™ Software v1.0 Staff Elimination Database Application
Applied Biosystems™
Applied Biosystems™ RapidLINK™ Software v1.0 Staff Elimination Database Application
Have Questions?
Catalog NumberQuantity
A418161 annual license
Catalog number A41816
Price (USD)
-
Quantity:
1 annual license
The Applied Biosystems RapidLINK Staff Elimination Database Application enables low-effort monitoring of possible contamination from laboratory and agency staff members, manufacturing employees, and other personnel who may come in contact with DNA during the process of generating a DNA profile.
The matching function between profiles in the RapidLINK database and the staff database runs in the background at all times, alerting the RapidLINK Software user only when there is a possible contamination event. The RapidLINK Staff Elimination Database Application automatically generates a printable summary report identifying the profile for further review and resolution. For security of the elimination database, only RapidLINK administrators with the proper access levels can upload employee profiles or remove inactive ones to keep the elimination database current. For use only with Applied Biosystems RapidLink Software v1.0.
Features of the RapidLINK Staff Elimination Database Application include:
• Identification of staff-related contamination
• Meets a lab quality requirement
• Reduced workload for lab management
• Background application is always “on”
• Automatically generated report
• Administrator-defined database
Specifications
For Use With (Equipment)Applied Biosystems RapidLINK Software V1.0
Product TypeStaff Elimination Database Application Software
Quantity1 annual license
Shipping ConditionRoom Temperature
Product LineRapidLINK
Software TypeLicense
Unit SizeEach
Contents & Storage
1 document letter