Applied Biosystems™

Applied Biosystems™ RapidHIT™ ID System Upgrade

Catalog number:  A41821

 Related applications:

Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Applied Biosystems RapidHIT ID System Upgrade gives IntegenX RapidHIT ID System owners the ability to upgrade their system to the latest Applied Biosystems-branded version. The upgrade provides improved system reliability and performance, and enables the system to be sample/data accessioning workflow-capable.

Before upgrading, please consume your remaining IntegenX RapidHIT ID consumables as several legacy consumables, such as the primary cartridges, are not compatible with the Applied Biosystems-branded RapidHIT ID System.

For more details or questions, please contact your local sales representative or local distributor.

For Forensic and Human Identity Applications Only.

Specifications

Product Line: RapidHIT™
Product Size: 1 upgrade
Shipping Condition: Room Temperature

Contents & storage

1 instrument packaging kit.

Store at room temperature.

Documents

Manuals & protocols