Product Image
Thermo Scientific™
3500/3500xL Data Collection Software, v3.3
This product is a software upgrade package. It includes a 3500/3500xL Data Collection Software upgrade CD for upgrade from v3 or v3.1 to v3.3.
Have Questions?
Catalog NumberQuantity
A418571 license
Catalog number A41857
Price (USD)
-
Quantity:
1 license
This product is a software upgrade package. It includes a 3500/3500xL Data Collection Software upgrade CD for upgrade from v3 or v3.1 to v3.3.
Data Collection Software v3.3 provides the features and functionality that researchers have been requesting to take their research even further. Powered by the Windows™ 10 operating system, our newest and most powerful data collection software provides:
  • Options to enable/disable consumable hard stop warnings
  • Option to enable/disable consumable hard stops for administrator
  • Signal Optimization Factor parameter added to spatial calibration
  • Bi-weekly maintenance option added to Scheduler
  • Ability to export consumable logs
  • Ability to export injection list
  • Ability to load another plate and re-order priorities after run is paused
  • Added run pause confirmation dialog
  • Support to load additional plate and re-order injections after run is paused
  • Ability to enable/disable off-scale recovery and pull-up reduction
  • Additional features include:
  • Support for Windows 10, 64 bit
  • Security—built-in SAE feature set assists in meeting more stringent requirements
  • Ability to upload sample files to Connect cloud platform in non-SAE mode
Specifications
Includes3500/3500xL Data Collection Software v3/v3.3 to v3.3 CD, 1 licensing kit with single license code, combined RUO and SAE (Security, Audit, and E-signature), 1 user document CD
Quantity1 license
For Use With (Application)Data Collection
For Use With (Equipment)3500 Genetic Analyzer
Operating SystemMicrosoft Windows 10 IoT 64-bit
Product TypeData Collection Software
Unit SizeEach
Frequently asked questions (FAQs)
How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?
Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.
Find additional tips, troubleshooting help, and resources within our Fragment Analysis Software Support Center.