BenchStable™ MEM
Gibco™

BenchStable™ MEM

Green features
Gibco Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
A419220210 x 500 mL
A4192201500 mL
카탈로그 번호 A4192202
제품 가격(KRW)
599,000
온라인 행사
Ends: 31-Dec-2026
665,000
할인액 66,000 (10%)
Each
수량:
10 x 500 mL
제품 가격(KRW)
599,000
온라인 행사
Ends: 31-Dec-2026
665,000
할인액 66,000 (10%)
Each

Gibco Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

BenchStable MEM has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional MEM when supplemented with ≥10% fetal bovine serum (FBS).

This MEM is modified as follows:
• Optimized for room temperature stability
• With GlutaMAX, Phenol Red
• Without HEPES

Using BenchStable MEM
MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This MEM includes Earle's salts. MEM with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable MEM requires supplementation with ≥10% fetal bovine serum (FBS).

For Research Use Only. Not for use in diagnostic procedures.
사양
세포주HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
농도1 X
그린 기능ACT Ecolabel (Gold)
제조 품질cGMP-compliant under the ISO 9001: 2015 standard
제품라인BenchStable
제품 유형MEM (Minimum Essential Medium)
수량10 x 500 mL
유통 기한12 Months From Date of Manufacture
배송 조건Room Temperature
분류Animal Origin-free
형태Liquid
Serum LevelStandard Serum Supplementation
멸균Sterile-filtered
첨가제 포함Low Glucose, GlutaMAX, Phenol Red
첨가제 없음No HEPES, No Sodium Pyruvate
Unit SizeEach
구성 및 보관
Store at room temperature. Protect from light.

자주 묻는 질문(FAQ)

Is it okay if I supplement BenchStable media with more than 10% FBS?

We recommended supplementing BenchStable media with at least 10% FBS for best results. If your culture method calls for greater than 10% FBS supplementation using our other existing media, we recommend that you use the same concentration with BenchStable media.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it okay if I supplement BenchStable media with 5% FBS?

We recommend supplementing BenchStable media with at least 10% FBS for best results. We have run tests on limited cell types using 5% FBS concentration and have not observed compromised performance. Please contact Tech Support at techsupport@thermofisher.com to ensure that your specific application allows for 5% serum supplementation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which cell lines have been tested in BenchStable media?

We have tested our BenchStable media for growth and morphology on the following cell lines: A549, CHO-K1, HEK293, HeLa, HL-60, Jurkat, MCF7, MDA-MB-231, Neuro2A, SH-SY5Y, and THP-1.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer BenchStable media without phenol red?

At this time, we do not offer BenchStable media without phenol red.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will the shelf-life of BenchStable media improve if stored at lower temperature (2-8 degrees C)?

The shelf life of BenchStable media is unchanged when stored cold (2-4 degrees C) or ambient (15-30 degrees C).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.