High-Intensity Perfusion CHO Medium, 370 L - FAQs
High-Intensity Perfusion CHO Medium, 370 L - FAQs
View additional product information for High-Intensity Perfusion CHO Medium - FAQs (A4230201, A4230204, A4230203, A4230202)
5 product FAQs found
What feeding supplementation do you recommend for High-Intensity Perfusion (HIP) CHO Medium?
In general, High-Intensity Perfusion (HIP) CHO Medium was not intended to be paired with concentrated feeds and as such may not benefit from them the way that other media intended for fed-batch application do.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can CHO cells that are created or frozen in other production media be thawed into High-Intensity Perfusion (HIP) CHO Medium or adapted for growth and production?
Cells that are grown in or adapted to, and frozen in Gibco media such as CD CHO, CH OptiCHO, FortiCHO, Dynamis, and ExpiCHO seem to readily adapt to High-Intensity Perfusion (HIP) CHO Medium. However, adaptation should always be first demonstrated in flasks as suggested in the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019412_High-IntensityPerfusionCHO_Medium_UG.pdf). Only when it is proven that a cell clone suffers no growth lag should it be thawed directly into HIP CHO Medium and/or switched into HIP CHO Medium mid-process workflow.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does High-Intensity Perfusion (HIP) CHO Medium require supplementation for use?
When using a CHO cell line with a selection system other than the glutamine synthetase (GS) system, we recommend adding 2-8 mM L-Glutamine or GlutaMAX Supplement to High-Intensity Perfusion (HIP) CHO Medium. Additionally, we recommend making sure that glucose levels do not fall below 2 g/L in culture by supplementing with glucose as needed. Also, if clumping becomes an issue, then anti-clumping agent (ACA) can be used.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How is High-Intensity Perfusion (HIP) CHO Medium different from fed-batch medium?
Fed-batch oriented formulations typically perform best when paired with a feed. High-Intensity Perfusion (HIP) CHO Medium, by comparison, is formulated at a higher native concentration and assumes constant medium exchange without the addition of other feeds. Further, the osmolality of HIP CHO Medium at standard concentration is much higher than that of a typical fed-batch medium with the understanding that typical perfusion results in a lower osmolality. Lastly, HIP CHO Medium supports being reconstituted at multiple concentrations to better support the wide array of perfusion operating environments and seed train vs production needs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
When should I consider using High-Intensity Perfusion (HIP) CHO Medium?
There are many situations where you may want to consider using High-Intensity Perfusion (HIP) CHO Medium:
- Want to switch to perfusion with transfected CHO suspension culture
- Want a single platform with minimal complexity
- Desire high process flexibility
Additionally, HIP CHO medium allows sustained continuous production perfusion for long duration.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.