CTS™ PSC Cryopreservation Kit
Gibco™

CTS™ PSC Cryopreservation Kit

The CTS PSC Cryopreservation Kit is cGMP manufactured and used for the cryopreservation and recovery of pluripotent stem cells (PSCs).
Have Questions?
Catalog NumberQuantity
A42393011 Kit
Catalog number A4239301
Price (MXN)
-
Quantity:
1 Kit

The CTS PSC Cryopreservation Kit is cGMP manufactured and used for the cryopreservation and recovery of pluripotent stem cells (PSCs). The kit consists of CTS PSC Cryomedium, a xeno-free solution for the cryopreservation of early passage PSCs, and CTS RevitaCell Supplement (100X), a chemically defined recovery supplement for use in the post-thaw culture medium. In combination, these reagents help minimize loss of cell viability, maximize post-thaw recovery, and minimize unwanted differentiation of pluripotent stem cells.

The CTS PSC Cryopreservation Kit:

  • Provides efficient recovery and higher viability than traditional methods of PSC cryopreservation
  • Facilitates regulatory filings through cGMP manufacturing and the availability of documentation
  • Simplifies the transition from bench to clinic through the direct replacement of the RUO PSC Cryopreservation Kit

Efficient recovery
The CTS PSC Cryopreservation Kit achieves higher viability than traditional methods of cryopreservation. It is compatible with clump and single cell passaged PSCs cultured in a variety of media systems. CTS PSC Cryomedium is provided as a ready-to-use, complete solution containing 10% DMSO for efficient cell addition. Separate additions of enhancing reagents are not required, helping save valuable preparation time.

Facilitates regulatory filings
The CTS PSC Cryopreservation Kit is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability and convenient access to our Drug Master File (DMF) are available for the CTS PSC Cryopreservation Kit.

Simplifies transition from bench to clinic
The CTS PSC Cryopreservation Kit provides the same efficient recovery, consistency, and reduced variability as the RUO PSC Cryopreservation Kit. With its additional safety testing, including adventitious agents, the CTS PSC Cryopreservation Kit enables a smooth transition from the RUO PSC Cryopreservation Kit.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Specifications
Cell TypeEmbryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBottle
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product TypeCryopreservation Medium
Quantity1 Kit
Shelf Life12 months from date of manufacture
Shipping ConditionCTS PSC Cryomedium: Dry Ice
CTS RevitaCell: Dry Ice
ClassificationXeno-free
FormLiquid
SterilitySterile-filtered
With AdditivesDMSO (10%)
Unit SizeEach
Contents & Storage
• 50 mL CTS PSC Cryomedium; store at –5 to –20°C and protect from light
• 5 mL CTS RevitaCell Supplement (100X); store at –5 to –20°C and protect from light

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

What is the difference between the PSC Cryopreservation Kit and the CTS PSC Cyropreservation Kit?

The CTS PSC Cryopreservation Kit (Cat. No. A4239301) provides the same efficient recovery, consistency, and reduced variability as the PSC Cryopreservation Kit (Cat. No. A2644601). However, these two kits have different intended use statements, i.e., “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals” and “For Research Use Only”, respectively. Furthermore, the CTS PSC Cryopreservation Kit has full USP sterility testing info and a Drug Master file (DMF), whereas the sterility testing of the PSC Cryopreservation is via a modified USP method, and it does not have a DMF.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.