Pharma KingFisher™ Flex 96 Deep-Well Tip Combs
Applied Biosystems™
Pharma KingFisher™ Flex 96 Deep-Well Tip Combs
These tip combs are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96Read more
Have Questions?
Catalog NumberQuantity
A43074100 Combs
Catalog number A43074
Price (USD)
870.00
Each
Quantity:
100 Combs
Price (USD)
870.00
Each
These tip combs are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.
Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KingFisher™ Flex
Product LineKingFisher Flex
Product TypeTip Comb
Quantity100 Combs
Shipping ConditionRoom Temperature
Unit SizeEach
Frequently asked questions (FAQs)
My Pharma KingFisher Flex run was stopped due to an error message. Can I start the protocol from where it stopped?
No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand, or a modified version of the original Pharma KingFisher Flex script would have to be created that starts off at the point where the run was stopped.
Is it always necessary to use the transport locks on the Pharma KingFisher Flex instrument?
The transport locks are only necessary when relocating the instrument.
Are the volumes of reagents in each well critical for a successful Pharma KingFisher Flex run?
We strongly recommend that you maintain the specified volumes within the defined limits to avoid spillover and to help ensure the best performance at the most efficient level.
What if the magnetic particles are attached to the tip combs after my Pharma KingFisher Flex run?
This happens sometimes but it will not affect the yield because the sample has been released from the particles.
What if the magnetic particles remain in the sample well during a Pharma KingFisher Flex run?
If the starting material is too viscous, the magnetic rods will not be able to collect the particles. Dilute the sample and check that the sample is properly homogenized/lysed. Adding low amounts of detergent will improve the collection of magnetic particles as well. Quickly centrifuge the plate to sediment the particles to the bottom of the plate.