Pharma KingFisher™ Flex 96 Deep-Well Plates
Applied Biosystems™
Pharma KingFisher™ Flex 96 Deep-Well Plates
These deep-well plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96Read more
Have Questions?
Catalog NumberQuantity
A4307550 plates
Catalog number A43075
Price (USD)
439.00
Each
Quantity:
50 plates
Price (USD)
439.00
Each
These deep-well plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits. The deep-well configuration differs from the standard configuration in that the volume of sample input is from 100 to 1,000 μL per well compared to 20–200 μL for the standard configuration.
Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KingFisher™ Flex
Packaging50 Plates
Product TypeMicroplate
Quantity50 plates
Shipping ConditionRoom Temperature
BarcodeNo Barcode
ColorClear
MaterialPolypropylene
No. of Wells96
Product LineKingFisher Flex
Unit SizeEach
Contents & Storage
50 deep-well 96-well plates
Frequently asked questions (FAQs)
My Pharma KingFisher Flex run was stopped due to an error message. Can I start the protocol from where it stopped?
No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand, or a modified version of the original Pharma KingFisher Flex script would have to be created that starts off at the point where the run was stopped.
Is it always necessary to use the transport locks on the Pharma KingFisher Flex instrument?
The transport locks are only necessary when relocating the instrument.
Are the volumes of reagents in each well critical for a successful Pharma KingFisher Flex run?
We strongly recommend that you maintain the specified volumes within the defined limits to avoid spillover and to help ensure the best performance at the most efficient level.
What if the magnetic particles are attached to the tip combs after my Pharma KingFisher Flex run?
This happens sometimes but it will not affect the yield because the sample has been released from the particles.
What if the magnetic particles remain in the sample well during a Pharma KingFisher Flex run?
If the starting material is too viscous, the magnetic rods will not be able to collect the particles. Dilute the sample and check that the sample is properly homogenized/lysed. Adding low amounts of detergent will improve the collection of magnetic particles as well. Quickly centrifuge the plate to sediment the particles to the bottom of the plate.