Attune™ NxT and CytPix Flow Cell Cleaning Solution
The quality of your flow cytometry data can be affected by the cleanliness of the flow cell. Attune NxT FlowRead more
Have Questions?
Catalog Number
Quantity
A43635
30 mL
Catalog number A43635
Price (USD)
36.65
Online exclusive
36.92
Each
Quantity:
30 mL
Price (USD)
36.65
Online exclusive
36.92
Each
The quality of your flow cytometry data can be affected by the cleanliness of the flow cell. Attune NxT Flow Cell Cleaning Solution is an alkaline liquid concentrate that, when mixed with water, removes contaminants without damaging the quartz glass used in the optical system of the Attune NxT Acoustic Focusing Cytometer.
Attune NxT Flow Cell Cleaning Solution has been formulated to minimize the contamination build-up that may occur in the system lines or flow cell after repeated usage. This solution is recommended for use during the SIP sanitize function just prior to the shutdown procedure. The recommended frequency of use depends upon the number of samples analyzed on the instrument. For high volume users, it is recommended as a daily preventive measure. For less frequent users, weekly use is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Block FormatNon-Interchangeable
For Use With (Equipment)Attune™ Acoustic Focusing Cytometer
Product LineAttune
Quantity30 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Product TypeCell Cleaning Solution
Unit SizeEach
Contents & Storage
Store at room temperature
Frequently asked questions (FAQs)
Why do I have to use ultra pure water for dilution of the Attune NxT Flow Cell Cleaning Solution?
The Attune NxT Flow Cell Cleaning Solution is filtered to remove particulates to avoid scratching optical surfaces, it is important not to introduce particulates from the water, hence, we recommend using ultra pure water for dilution.
How do I use the Attune NxT Flow Cell Cleaning Solution?
The directions for use are provided on the vial label: Dilute 1:3 in ultra pure water. Use the solution with the SIP sanitize protocol just prior to each shutdown every day.