TaqMan™ MON89788 Quantification Kit
Thermo Scientific™

TaqMan™ MON89788 Quantification Kit

The Thermo Scientific™ TaqMan™ MON89788 Quantification Kit enables relative quantification of as little as 0.1% of MON89788 (Roundup Ready™ 2) transgenic soybeans with respect to total soya in a sample. The kit is ISO 9001 and 14001 compliant.

Have Questions?
Catalog NumberQuantity
A441131 Kit
Catalog number A44113
Price (USD)
-
Quantity:
1 Kit

The Thermo Scientific™ TaqMan™ MON89788 Quantification Kit enables relative quantification of as little as 0.1% of MON89788 (Roundup Ready™ 2) transgenic soybeans with respect to total soya in a sample. The kit is ISO 9001 and 14001 compliant.

The absolute limit of quantification is 20 copies of DNA, and the detection limit of the PCR technique is 5 copies each of MON89788 and soya DNA.

The TaqMan™ MON89788 Quantification Kit includes:

  • Primers and TaqMan™ probes for real-time PCR detection of:
    • The transgenic event characteristic of the soybean line MON89788
    • The endogenous soy gene, lectin
  • MON89788 Standard, a plasmid DNA quantitation standard containing equimolar quantities of both MON89788 and lectin targets (1:1 ratio)
  • Enzyme and other buffer components necessary for real-time PCR

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast real-time PCR systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a Suite of Solutions For GMO Detection and Quantification

The TaqMan™ MON89788 Quantification Kit is just one of our many available kits for the sensitive detection of GMO meat species in food and feed samples.

Specifications
Detection MethodPrimer-probe
No. of Reactions50
Quantity1 Kit
Sample TypePlant, food, and feed
Storage Requirements

MON89788 Master Mix (store at -20°C) Soya Master Mix (store at -20°C)General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) MON89788 Standard (store at -20°C)

Target GeneMON89788 transgenic soybean DNA
Target Specificity0.1% of MON89788 transgenic soybeans with respect to total soya in a sample
For Use With (Application)Food Quality Testing
Unit SizeEach