Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL
Thermo Scientific™

Zeba™ Dye and Biotin Removal Spin Columns and Filter Plates, 0.5–10 mL

Enable fast and efficient removal of non-reacted fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples with Thermo Scientific Pierce Zeba Dye and Biotin Removal columns and filter plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatVolume (Metric)
A443042 Plates96-well Filter Plate96-well
A44296S5 ColumnsSpin Column0.5 mL
A4429625 ColumnsSpin Column0.5 mL
A442972 x 25 ColumnsSpin Column0.5 mL
A442985 ColumnsSpin Column2 mL
A4429925 ColumnsSpin Column2 mL
A443005 ColumnsSpin Column5 mL
A4430125 ColumnsSpin Column5 mL
A443025 ColumnsSpin Column10 mL
A443035 x 5 ColumnsSpin Column10 mL
A443052 x 2 Plates96-well Filter Plate96-well
Catalog number A44304
Price (USD)
440.65
Online exclusive
474.00
Save 33.35
Each
Quantity:
2 Plates
Format:
96-well Filter Plate
Volume (Metric):
96-well
Price (USD)
440.65
Online exclusive
474.00
Save 33.35
Each
Quickly remove non-reacted fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples with a 7K MWCO in fewer than 15 minutes with Thermo Scientific Pierce Zeba Dye and Biotin Removal columns and filter plates.

Removing free-dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. Pierce Zeba Dye and Biotin Removal resin is highly specialized to produce exceptional protein recoveries while effectively removing non-conjugated fluorescent dyes, biotinylation reagents, reducing agents, and crosslinkers. When using the appropriate amount of sample and working within buffer conditions, these columns and filter plates can successfully remove almost any fluorescent dye. Even dilute protein samples (25 μg/mL) can be successfully processed to remove dyes, biotin, reducing agents, and crosslinkers and to efficiently recover proteins (>7 kDa). These products are not recommended for desalting or buffer exchange.

Features of Zeba Dye and Biotin Removal columns and filter plates include:
• Versatile—removes unreacted fluorescent dyes, biotinylation reagents, crosslinkers, and reducing agents from protein solutions
• High recovery—low-binding resin maximizes protein recovery
• Easy-to-use—no cumbersome column preparation or equilibration
• Fast—small-molecule removal and protein recovery in less than 15 minutes
• Flexible—available in spin columns and filter spin plates for a range of needs

Sample capacities
• 0.5-mL spin column: 50 to 120 μL sample
• 2-mL spin column: 400 to 700 μL sample
• 5-mL spin column: 1 to 2 mL sample
• 96-well filter plate: 40 to 100 μL sample (per well)

Available formats
• Spin columns: 0.5, 2, 5, and 10 mL
• 96-well filter plates: pre-dispensed; compatible with centrifugation for manual or automated purification; enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput clean-ups

Applications
• Ideal for post-reaction clean-up of samples
• To effectively remove non-conjugated dye, biotin, reducing agents, and crosslinkers with exceptional protein recovery

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2 Plates
Shipping ConditionRoom Temperature
Volume (Metric)96-well
Column TypeProprietary Resin
Format96-well Filter Plate
Product LineZeba
Sample Volume (Metric)40 to 100 μL
Unit SizeEach
Contents & Storage
Store at 4°C.

Frequently asked questions (FAQs)

Are Zeba Dye and Biotin Removal Spin Columns/Filter Plates compatible with mass spectrometry?

Zeba Dye and Biotin Removal Spin Columns/Filter Plates are not directly compatible with mass spectrometry.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Have you tested the Zeba Dye and Biotin Removal Spin Columns/Filter Plates using fixed angle rotor and swinging bucket rotor centrifuges?

The performance of the Zeba Dye and Biotin Removal resin is not affected by the type of centrifuge rotor.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How dilute can my protein sample be for using with Zeba Dye and Biotin Removal Spin Columns/Filter Plates?

Protein samples with as low a concentration as 25 µg/mL can be successfully processed with Zeba Dye and Biotin Removal Spin Columns/Filter Plates to remove dyes, biotin, reducing agents, and crosslinkers and to efficiently recover proteins (>7 kDa).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

When should I use Zeba Dye and Biotin Removal Spin Columns/Filter Plates?

Zeba Dye and Biotin Removal Spin Columns/Filter Plates enable fast and efficient removal of non-conjugated fluorescent dyes, biotin, reducing agents, and crosslinkers from protein samples. We have tested removal of the following:
- Alexa Fluor dyes: 488, 555, 647
- Dylight dyes: 488, 550, 650, 650-4x PEG
- Rhodamine dye
- Biotin
- Reducing agents: DTT, TCEP
- Crosslinker: BS3

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.