MycoSEQ™ Assay Service
Product Images
Thermo Scientific™
MycoSEQ™ Assay Service
MycoSEQ Assay Service is a fast and cost-effective way to verify the absence of mycoplasma in your cell lines. ThisRead more
Have Questions?
Catalog number A44316
Price (USD)/ 1 sample
197.00
-
Add to cart
Price (USD)/ 1 sample
197.00
Add to cart
MycoSEQ™ Assay Service
Catalog numberA44316
Price (USD)/ 1 sample
197.00
-
Add to cart
MycoSEQ Assay Service is a fast and cost-effective way to verify the absence of mycoplasma in your cell lines. This service leverages the MycoSEQ Mycoplasma Detection Kit, a real-time PCR assay designed and validated to meet the requirements of European Pharmacopeia Chapter 2.6.7. The primers were designed using a proprietary bioinformatics pipeline to detect >90 mycoplasma species with high specificity and no cross-reactivity to closely related bacterial species.
The MycoSEQ Assay Service offers:
  • Fast data delivery – turnaround time of four weeks or less
  • Convenient sample prep – just send a frozen cell pellet
  • High sensitivity – demonstrated by internal and external validations to detect 10 cfu/mL
  • High specificity – includes proprietary Discriminatory Positive Control to differentiate true positive from any potential cross-contamination positives
  • Exceptional support – a dedicated project manager for frequent project communication
How the MycoSEQ Assay Service works:
What you get
You will receive a comprehensive report that includes a data table confirming the results for your samples alongside results for the positive and negative assay controls.
Specifications
Quantity1 sample
Duration3 Weeks
Product TypeAssay Service
Product LineApplied Biosystems™, QuantStudio™
FieldResearch Use Only
Target AudienceCell Line Developers
SpeciesAll
Unit Size1 sample