Commercial sale and application support for this item is provided by Floragenex, our global authorized distributor. Please contact Floragenex at agriseq.orders@floragenex.com for purchasing, pricing, and product inquiries.
IonCode™ Barcode Adapters, 1153-1536
Ion Torrent™
IonCode™ Barcode Adapters, 1153-1536
IonCode Barcode Adapters 1153-1536 are 384 unique barcode adapters designed for optimal performance with Ion Torrent semiconductor sequencers. This setRead more
Have Questions?
Catalog NumberQuantity
A4460325 μL (each barcode)
Catalog number A44603
Price (USD)
-
Quantity:
25 μL (each barcode)
IonCode Barcode Adapters 1153-1536 are 384 unique barcode adapters designed for optimal performance with Ion Torrent semiconductor sequencers. This set includes IonCode Barcode Adapters 1301-1396, 1401-1496, 1501-1596, and 1601-1696. When used in combination with other IonCode barcode adapters and the AgriSeq HTS Library Kit, users can pool up to 1152 amplicon libraries and then conduct multiplexed sequencing analysis, which helps increase lab efficiency and reduce sequencing costs per sample.
Key product features include:
• Robust barcodes ligate efficiently and uniformly to generate balanced representation of libraries
• Unbiased performance enables identical genotypes independent of barcode used
• Robust error correction built in by design for added confidence in sample identification
Increase sample multiplexing without impacting data quality
Multiplexing with barcoded libraries enables more cost effective runs by permitting up to 1152 samples per run and helps significantly decrease the cost and handling requirements of sequencing. Each barcode was individually evaluated to verify no bias of sequence context representation. The barcodes were also screened for equivalent efficiency to verify balanced read depth between libraries. Regardless of the specific barcode used, AgriSeq targeted genotyping-by-sequencing (GBS) generates highly uniform libraries and concordant genotype calls.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion Chef™ System, Ion GeneStudio S5 System, Ion Proton™ System
LibrariesTargeted Sequencing Library
Product LineIonCode
Quantity25 μL (each barcode)
Shipping ConditionDry Ice
Workflow StepLibrary Barcoding
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Frequently asked questions (FAQs)
What precautions should I take while handling IonCode Barcode Adapters, 1153-1536?
When handling IonCode barcode adapters, be careful to avoid cross‑contamination by changing gloves frequently.
- Pulse vortex the IonCode Barcode Adapters plate several times.
- Centrifuge the IonCode Barcode Adapters plate for 1 min at 1000 x g to collect contents at the bottom of the wells.
- Carefully remove and then discard the plate seal.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the shipping and storage conditions for IonCode Barcode Adapters, 1153-1536?
IonCode Barcode Adapters, 1153-1536 is shipped on dry ice, and we recommend storing at -30 degrees C to -5 degrees C, in an upright position.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How are IonCode Barcode Adapters, 1153-1536 supplied?
IonCode Barcode Adapters, 1153-1536 are supplied in 4 separate 96-well plates containing 25 µL of each barcode:
- IonCode Barcode Adapters 1301-1396 in 96-well PCR plate (red)
- IonCode Barcode Adapters 1401-1496 in 96-well PCR plate (yellow)
- IonCode Barcode Adapters 1501-1596 in 96-well PCR plate (green)
- IonCode Barcode Adapters 1601-1696 in 96-well PCR plate (blue)
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are IonCode Barcode Adapters, 1153-1536?
IonCode Barcode Adapters, 1153-1536 are 384 unique barcode adapters designed for optimal performance with Ion Torrent semiconductor sequencers. This set includes IonCode Barcode Adapters 1301-1396, 1401-1496, 1501-1596, and 1601-1696.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What can I use IonCode Barcode Adapters, 1153-1536 for?
When used in combination with other IonCode barcode adapters (Cat. Nos. A31173, A36546, and A44599) and the AgriSeq HTS Library Kit (Cat. Nos. A34143 and A34144), IonCode Barcode Adapters, 1153-1536 enables pooling up to 1,536 amplicon libraries to conduct multiplexed sequencing analysis.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.