Complete Raman Library Collection
Thermo Scientific™
Complete Raman Library Collection
The Complete Raman Library Collection includes unlimited access for one user, for one full year, to more than 17,000 high-quality raman spectra in high-resolution format.
Have Questions?
Número de catálogoTipo
A45136Biblioteca espectral
Número de catálogo A45136
Precio (MXN)
-
Tipo:
Biblioteca espectral
The Complete Raman Library Collection includes unlimited access for one user, for one full year, to more than 17,000 high-quality raman spectra in high-resolution format.
All libraries can be accessed using the search functionality included in the OMNIC Anywhere application available through our Connect cloud-based platform. Using data in your Connect account, your OMNIC data files can be quickly searched and results can be downloaded in several different file and report formats.
Subscriptions are purchased through ThermoFisher.com and delivered/managed through your Connect account. You will be notified of any changes to your subscription, including upcoming expiration dates.
Library updates are included with your subscription, so you will always have access to the latest spectral data.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Línea de productosThermo Scientific
Cantidad1 user, 1 year
TipoBiblioteca espectral
Unit SizeEach