Genexus™ Cell-Free Total Nucleic Acid Purification Kit
Ion Torrent™

Genexus™ Cell-Free Total Nucleic Acid Purification Kit

The Ion Torrent Genexus Cell-Free Total Nucleic Acid Purification Kit for the Genexus Purification System is a set of consumablesRead more
Have Questions?
Catalog NumberNo. of Reactions
A4554224
Catalog number A45542
Price (MXN)
16,886.27
Each
No. of Reactions:
24
Price (MXN)
16,886.27
Each
The Ion Torrent Genexus Cell-Free Total Nucleic Acid Purification Kit for the Genexus Purification System is a set of consumables that enables automated extraction and quantitation of TNA from cell-free plasma, isolated from whole blood. This kit supports four runs of up to six samples each, utilizing the proven MagMAX technology for extraction and Qubit technology for post-extraction DNA quantitation.

Features of the Genexus Cell-Free Total Nucleic Acid Purification Kit include:
• Achieve consistent extraction results with MagMAX technology
• Reduce hands-on setup time using pre-filled consumables
• Maximize efficiency with automated Qubit quantitation
• Experience rapid turnaround times of as little as 3.25 hours from lysate to quantified nucleic acid

Prefilled reagents
The Genexus Purification System consumables are prefilled with most reagents necessary to perform the extraction, so only a few pipetting steps are required per sample to set up the run. Simply remove the foil seal, add binding buffer, enzymes, and your samples, then place the consumables onto the instrument.

Two output types: archive plate and NGS-ready output plate
After extraction, all nucleic acids are transferred directly to an archive plate and kept cool for up to one hour after run completion. The 48‐well nucleic acid archive plate is barcoded to facilitate tracking of sample extractions across several stored archive plates.

When the Genexus Purification System is connected to the Genexus Integrated Sequencer and run as part of an integrated workflow, a predefined aliquot of each extracted nucleic acid is transferred from the archive plate to an NGS-ready 96‐well nucleic acid output plate, which serves as the sample input to the Genexus Integrated Sequencer.

For Research Use Only. Not for use in Diagnostic Procedures.
Specifications
For Use With (Application)Next Generation Sequencing
For Use With (Equipment)Genexus™ Purification System
No. of Reactions24
OutputPurified Total Nucleic Acid (TNA), Quantified DNA
Purification MethodMagMax™ bead-based purification
Sample TypeCell-free Plasma (Isolated from Whole Blood)
Shipping ConditionAmbient
Workflow StepSample Purification and Quantification
FormatPre-filled Reagent Plates
Unit SizeEach