EasyPep™ MS Sample Prep Kits
EasyPep™ MS Sample Prep Kits
EasyPep™ MS Sample Prep Kits
Thermo Scientific™

EasyPep™ MS Sample Prep Kits

Generate high quality proteomic samples for MS analysis in less than four hours using optimized EasyPep MS Sample Prep kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
A45735Lysis Buffer
A40006Mini MS Sample Prep Kit
A57864Micro MS Sample Prep Kit
A45733MS Sample Prep Kit
A45734Maxi Sample Prep Kit
A57865Peptide Clean-up Plate
Catalog number A45735
Price (USD)
130.65
Online exclusive
150.00
Save 19.35
Each
Description:
Lysis Buffer
Price (USD)
130.65
Online exclusive
150.00
Save 19.35
Each

EasyPep Mini MS Sample Prep Kits enable efficient and reproducible processing of cultured mammalian cells and tissues for proteomic MS analysis. The kit contains pre-formulated buffers, MS-grade enzyme mix, peptide clean-up columns, and an optimized protocol to generate MS-compatible peptide samples in less than three hours. Kits are optimized to process protein samples from 10–100 μg with a high yield of MS-ready peptides.

• Complete—includes pre-formulated reagents for lysis through digestion, peptide clean-up columns, and an optimized protocol for up to 20 samples
• Optimized—streamlined protocol and reagents minimize the number of steps and time time needed to process samples
• Flexible—reagents and protocol have been verified using cells, plasma, and tissue samples for 10 to 100 μg samples
• Time-saving—sample processing time reduced from more than one day to less than three hours
• Compatible—final preparation is ready for direct MS analysis and other downstream applications, including TMT labeling

The sample preparation of peptides for MS analysis is complex, with numerous steps and varied protocols, resulting in sample variability in both quality and quantity. This kit has been designed to improve reproducibility while saving hands-on and processing time. The number of steps and processing time have been reduced through the addition of Universal Nuclease to reduce viscosity from nucleic acids (replacing sonication), a rapid 'one pot' reduction/alkylation solution for cysteine modification (carbamidomethylation, + 57.02), and a trypsin/Lys-C protease mix for more complete digestion. In addition, the kit includes Peptide Clean-up columns and buffers to prepare detergent-free peptide samples for MS analysis. The optimized reagents and protocol produce high-quality peptides that are compatible with TMT labeling and other downstream applications, or are ready for MS analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionLysis Buffer
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeUnlabeled
Quantity100 mL
Shipping ConditionWet Ice
Workflow StepCell Lysis
Detection MethodMass Spectrometry
FormLiquid
Product LineEasyPep
Product TypeLysis Buffer
Starting MaterialProtein Samples
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C

Frequently asked questions (FAQs)

After cell lysis, my mass spectrometry sample is very viscous and difficult to pipette. How do I reduce the sample viscosity?

High sample viscosity after lysis is due to release of DNA from the nucleus. Sonication or addition of a nuclease such as the Pierce Universal Nuclease (Cat. No. 88700, 88701, or 88702) can be used to degrade DNA and reduce sample viscosity.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

How do I process yeast samples with EasyPep Mini MS Sample prep Kit?

Add 0.5 mm diameter glass beads to the sample pelleted in Easy Pep Lysis Solution and proceed as recommended in the Easy Pep sample prep protocol. Vortex vigorously for few minutes, spin at 16,000 x g for 5 mins, and extract the supernatant for protein assay and further downstream sample preparation.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

How do I process bacterial samples with EasyPep MS Sample Prep kits?

Add 1-2 µL of 50 mg/mL Lysozyme Solution (Cat. No. 90082) to 200 µL of the EasyPep lysis buffer provided in the kit and proceed with the sample prep. Add 200 µL of the prepared lysis solution to 5 X 10E8 bacterial cells (E. coli cells) (OD600 equal to 1, corresponding to 1 X 10E8 CFUs). Lyse by pipetting the sample repeatedly and centrifuge at 16,000 x g for 5 mins. Alternatively, E. coli cells can be lysed in lysis buffer using bead beating or sonication instead of lysozyme. After lysis, dilute samples to 1 mg/mL using lysis buffer, proceed with the EasyPep protocol for reduction, alkylation, digestion, and clean up as described in the product manual.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Do you offer a mass spec sample prep method for low abundant proteins in samples?

High Select Top14 Abundant Protein Depletion Mini Spin columns (Cat. Nos. A36369, A36370) can be used to deplete top 14 abundant plasma/serum proteins. The depletion of highly abundant proteins from the sample enables the detection and identification of low abundant proteins. The depleted samples can be processed with EasyPep MS sample prep kit to generate digested plasma/serum samples. The clean digested samples can be processed by nanoLC-MS/MS analysis for discovery experiments or nanoLC-PRM/MS analysis for targeted experiments.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

At which step should I label my mass spec samples with TMT reagents?

TMT labeling can be performed after reduction, alkylation, digestion, or prior to cleanup, if the sample is buffered at appropriate pH (8-8.5) in a suitable buffer without primary amines (e.g., Tris, glycine). TMT labeling can also be performed after peptide clean-up. Labeling of peptides after clean-up enables measuring and normalizing of the peptide samples for equal mixing.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.