Search
Search

| Catalog Number | Type | Warranty |
|---|---|---|
| A45927 | Quantitation Starter Kit | 3-year AB Repair Center Warranty |
| A44984 | Benchtop Fluorometer | 3-year Rapid Exchange Warranty |
| A44985 | Benchtop Fluorometer | 3-year AB Repair Center Warranty |
| Q33327 | Fluorometer | Standard Warranty |
| A45925 | Quantitation Starter Kit | 3-year Rapid Exchange Warranty |
| Q45894 | Quantitation Starter Kit | Standard Warranty |
| A45924 | NGS Starter Kit | 3-year Rapid Exchange Warranty |
| A45926 | NGS Starter Kit | 3-year AB Repair Center Warranty |
| Q45893 | NGS Starter Kit | Standard Warranty |
The Qubit Flex Fluorometer is designed for highly accurate and sensitive quantification of DNA, RNA, microRNA, protein, and endotoxin. This benchtop fluorometer offers the flexibility to directly measure the fluorescence of up to eight samples simultaneously, which reduces assay variability. Paired with Qubit reagents, the Qubit Flex Fluorometer is part of an optimized workflow generating highly accurate and reproducible results. The fluorometer has a large screen and intuitive user interface, making it easy to select and run your desired assay with results displayed in just a few seconds.
Features of the Qubit Flex Fluorometer include:
Higher throughput, less variability than single sample readers
The Qubit Flex Fluorometer has the flexibility to measure multiple DNA, RNA, protein, and endotoxin samples up to 50% faster than single-sample readers. In addition to saving time, the ability to measure eight samples simultaneously reduces variability, resulting in highly reproducible data. The fluorometer includes reagent, range, and molarity calculators, further reducing the time to results. The reagent and range calculators help to determine which reagent and what amount of sample to use to effectively quantify the DNA, RNA, or protein. The molarity calculator allows you to calculate the molarity of your samples based on nucleic acid length and the measured concentration.
Enables more accurate results than UV absorbance at low concentrations
The Qubit Flex Fluorometer is far more sensitive than UV absorbance, which measures anything absorbing at 260 nm, including DNA, RNA, protein, free nucleotides, and excess salts. Moreover, UV spectrophotometry is often not sensitive enough to accurately measure low concentrations of DNA and RNA. With the Qubit Flex Fluorometer, your research is enhanced by more accurate measurements because the dyes in the Qubit assay kits fluoresce only when bound to the target molecule (DNA, RNA, protein, or endotoxin) in your sample. This allows you to avoid repeating work due to inaccurate measurements.
Comprehensive warranty options
The Qubit Flex Fluorometer comes with a standard 1-year warranty that covers repair costs including shipping, parts, and labor. Service plans can be bundled with new instrument purchases to provide a 3-year warranty. These Qubit Flex Fluorometer packages are offered with two different types of bundled warranties:
21 CFR Part 11 Software Module
The Qubit Flex Security Software supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using the SAE administration console. The 21 CFR Part 11 software can be purchased separately (Cat. No. Q31994).