Product Image
Applied Biosystems™
3500 Series HID Data Collection Software v4.0.1
Applied Biosystems 3500 Series Data Collection Software, version 4.0.1, is designed for data collection in human identification (HID) applications andWeitere Informationen
Have Questions?
KatalognummerMenge
A460851 Lizenz
Katalognummer A46085
Preis (EUR)
-
Menge:
1 Lizenz
Applied Biosystems 3500 Series Data Collection Software, version 4.0.1, is designed for data collection in human identification (HID) applications and intended for use with the 3500 Series Genetic Analyzer. This product is also a software upgrade package. It includes a 3500/3500xL Data Collection Software upgrade CD for upgrade from v3.1 or v4.0 to v4.0.1.
Compatible operating system: Windows 10, 64-bit only
3500/3500xL Data Collection Software v4.0.1 provides:
• Reduced spectral pull-up for Thermo Fisher Scientific dye sets
• Off-scale recovery for Thermo Fisher Scientific dye sets
• New features to support off-scale recovery and reduced spectral pull-up
• Ability to set pre-expiry and expiry warning independently
• Signal Optimization Factor parameter added to 3500xL spatial calibration
• Size Standard Normalization and Average Normalization parameters added to the result view
• Bi-weekly maintenance option added to Scheduler
• Support for J6T dye set
• Off-scale recovery support for new dye sets
• Ability to export consumable logs
• Ability to export injection list
• Installation check support for HID 6-dye
• Support to load additional plate and re-order injections after run is paused
• Ability to enable/disable off-scale recovery and pull-up reduction
• Removal of Flexnet licensing
• Security—built-in SAE feature set assists in meeting more stringent requirements
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
Zur Verwendung mit (Geräte)3500 Genetic Analyzer
Produkttyp3500 Series HID Data Collection Software V4.0.1
Menge1 Lizenz
VersandbedingungRaumtemperatur
Software TypeCD
Unit SizeEach
Inhalt und Lagerung
Data Collection Software for HID on CD for use on 3500 Genetic Analyzer