PowerTrack™ SYBR Green Master Mix for qPCR
Applied Biosystems™

PowerTrack™ SYBR Green Master Mix for qPCR

PowerTrack SYBR Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows with built-in visual dye–based indicators for accurate reaction set up.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A4611350 mL
A460121 mL
A461095 mL
A461102 x 5 mL
A461115 x 5 mL
A4611210 x 5 mL
Catalog number A46113
Price (USD)
4,306.65
Online exclusive
4,785.00
Save 478.35
Each
Quantity:
50 mL
Price (USD)
4,306.65
Online exclusive
4,785.00
Save 478.35
Each

PowerTrack SYBR Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows with built-in visual dye–based indicators for accurate reaction set up. Coupled with user-supplied primer sets and template, PowerTrack SYBR Green Master Mix is designed to amplifiy targets for accurate gene expression analysis.

Features of PowerTrack SYBR Green Master Mix include:

  • Built-in visual indicators aid in reaction setup
  • Broad primer Tm and concentration compatibility allows qPCR reaction setup flexibility with minimal optimization
  • Specific and tight reproducibility in Ct's over a broad dynamic range
  • Formulated with UNG/dUTP to prevent contamination of carryover PCR products

Built-in visual indicators

PowerTrack SYBR Green Master Mix contains an inert blue dye that does not interfere with real-time PCR but provides increased visibility in the tube or well. A separate sample buffer is provided that contains an inert yellow dye. The optional use of this yellow sample buffer results in a reaction mixture that changes from blue to green upon addition of the sample, helping one avoid pipetting mistakes. The yellow sample buffer aids in reaction setup, but is not required to obtain superior results with PowerTrack SYBR Green Master Mix.

Formulated for maximum specificity and reproducibility

PowerTrack SYBR Green Master Mix contains Taq DNA polymerase, which is tightly controlled through an antibody-mediated hot-start mechanism, helping prevent undesirable early activity of the polymerase at low temperatures that can lead to nonspecific amplification. PowerTrack SYBR Green Master Mix is designed for exceptional performance in your most challenging real-time PCR applications.

Broad instrument compatibility

PowerTrack SYBR Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems real-time PCR instruments. It is also compatible with the Bio-Rad CFX96™, CFX384™, and iQ™5; Roche LightCycler™ 480; and Agilent Mx3005P™ systems. For optimal results, primer concentrations should be in the range of 300–800 nM.

UNG for carryover contamination control

Contamination in labs that routinely run PCR is a major concern for many researchers and is the source of most false positives. The inclusion of UNG and dUTP in PowerTrack SYBR Green Master Mix results in the degradation of any previously amplified PCR products and helps prevent contamination of subsequent qPCR reactions.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Flex, QuantStudio 7 Pro, QuantStudio 12K Flex, Bio-Rad CFX Connect, Bio-Rad CFX384 Touch, Bio-Rad CFX96 Touch, BioRad CFX96, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, ViiA 7 System, StepOne System, MJ Chromo4, MJ Opticon, StepOnePlus System, Roche LightCycler 480, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
FormatTube
IncludesdUTP, UNG/UDG
Inhibitor Tolerance (Simple)High
Multiplex CapabilitySingle-plex
No. of Reactions5000 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseTaq DNA Polymerase
Product LineApplied Biosystems
Product TypeReal Time PCR SYBR Master Mix
Quantity50 mL
Sample TypeDNA
Shipping ConditionDry Ice
Sufficient For5000 Reactions
Target SpecificitycDNA or gDNA only
Thermostability8 hr., Assembled Reaction
Volume1 x 50 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression, Microbial Detection
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
• PowerTrack SYBR Green Master Mix
• 40X Yellow Sample Buffer

Store at -20±5°C in the dark.

Frequently asked questions (FAQs)

What products do you recommend for downstream analysis by qPCR when using the SuperScript IV Single Cell/Low Input cDNA PreAmp Kit?

We recommend using TaqMan Fast Advanced Master Mix for all TaqMan‑based detection methods.

We recommend using PowerTrack SYBR Green Master Mix for all SYBR Green‑based detection methods.

When using the PowerTrack SYBR Green Master Mix, can I use cDNA premixed with yellow sample buffer?

We haven't tested this extensively, but we have tested storage of the cDNA and yellow sample buffer premixed for 2-3 days. For best practice, we recommend keeping the cDNA and yellow sample buffer separate until the reaction set up.

For the PowerTrack SYBR Green Master Mix, there is no description in the quick reference or user guide for UNG incubation time. What is the run time and temperature for the UNG incubation?

The incubation time is not required as UNG is active at room temperature and will degrade amplicons with dUs present.

Is there a specific reason why the PowerTrack SYBR Green Master Mix contains the SYBR Green I dye rather than SYBR GreenER dye?

SYBR Green I dye has been used in PowerTrack SYBR Green Master Mix because our product development team found the SYBR Green I dye to perform more optimally than SYBR GreenER dye during the development of this master mix.

What is the benchtop stability of PowerTrack SYBR Green Master Mix?

Pre-assembled qPCR reactions with PowerTrack SYBR Green Master Mix are stable for up to 8 hrs at room temperature before being run, when stored away from light.