Ion AmpliSeq™ Microbiome Health Research Kit
Ion Torrent™
Ion AmpliSeq™ Microbiome Health Research Kit
The Ion AmpliSeq Microbiome Health Research Kit is a targeted NGS assay for characterizing the human gut microbiome, which has been implicated in human disease.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoN.º de muestras
A46497384 muestras
A4649548 muestras
A46496256 muestras
Número de catálogo A46497
Precio (MXN)
-
N.º de muestras:
384 muestras
This product is available in different configurations, ranging from 48 up to 384 samples per kit to accommodate your Microbiome research needs. The product bundles provide reagents and materials for a cost-effective and comprehensive, targeted next-generation sequencing (NGS) workflow for highly sensitive, specific, and robust characterization of human gut microbiome from stool samples, with a full analysis solution providing optimized informatics and visualization.
The Ion AmpliSeq Microbiome Health Research Kit is a targeted NGS assay for characterizing the human gut microbiome, which has been implicated in human disease. The assay consists of two pools of Ion AmpliSeq oligonucleotide primers and associated reagents to generate amplicon libraries for NGS on Ion Torrent platforms. The assay targets multiple hypervariable regions in the 16S rRNA gene, as well as 73 species-specific sequences. With this kit, DNA isolated from human stool samples is used to generate libraries for templating and sequencing on the Ion S5 and Ion Chef systems.
Features of the Ion AmpliSeq Microbiome Health Research Kit include:
  • Simple – full sample-to-answer solution consisting of an easy and robust workflow including analysis software
  • Robust – enables profiling of the human gut microbiome with excellent sensitivity, specificity, and reproducibility
  • Comprehensive – enables accurate taxonomic profiling and relative abundance estimation targeting eight of nine 16S hypervariable regions of 16S rRNA gene and accurate detection of 73 key bacterial species associated with human disease
  • Efficient – enables screening of 32 samples multiplexed on a single Ion 540 Chip or 48 samples on a single In 550 Chip, with entire workflow typically completed in three days, including analysis
  • Sensitive – compatible with as little as 1 ng input DNA for 16S rRNA Gene Pool and 10 ng for Target Species Pool
This assay is designed to assist researchers in understanding the impact of gut microbiome diversity and composition on human health and disease. The kit enables global characterization of the microbiome by assaying multiple hypervariable regions in the 16S rRNA gene and detection of 73 key bacterial species associated with human diseases, such as cancer, including bacteria linked to potential efficacy of immune-modulatory drugs, gastrointestinal and immunological disorders, as well as infectious disease. We have designed species-specific primers that provide high strain coverage and high specificity performance using the best-in-class amplicon technology. With this approach, reads generated from sequencing can be analyzed to report taxonomic classifications and relative abundance for organisms in the sample with high specificity and sensitivity.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Método de detecciónNext Generation Sequencing
Para utilizar con (aplicación)Secuenciación
N.º de muestras384 muestras
Línea de productosIon AmpliSeq
Tipo de productoMicrobiome Health Research Kit
Cantidad384 samples
Condiciones de envíoDry Ice
EspecieHumano
FormatoKit
Unit SizeEach
Contenido y almacenamiento
• Ion AmpliSeq Microbiome Health Research Panel, store at -5 to -30°C
• Ion AmpliSeq Library Kit Plus (96 reactions), store at -5 to -30°C
• IonCode Barcode Adapters 0101–0196, store at -5 to -30°C
Catalog number A46496 also include:
• Ion 540 Chip Kit, store at room temperature
• Ion S5 Chef Solutions, store at room temperature
• Ion S5 Chef Supplies, store at room temperature
Catalog number A46497 also include:
• Ion 550 Chef Reagents, store at -5 to -30°C
• Ion S5 Sequencing Solutions, store at room temperature
• Ion S5 Sequencing Reagents, store at -5 to -30°C
Preguntas frecuentes
What is included in the Ion AmpliSeq Microbiome Health Research Kit, Ion 550 bundle?
The Ion AmpliSeq Microbiome Health Research Kit, Ion 550 bundle (Cat. No. A46497) consists of the Ion AmpliSeq Microbiome Health Research - 16S rRNA Gene Pool, the Ion AmpliSeq Microbiome Health Research - Target Species Pool, the Ion AmpliSeq Library Kit Plus, the IonCode Barcode Adapters, the Ion 550 Kit - Chef, and the Ion 550 Chip Kit for the rapid preparation of barcoded sample libraries from bacterial DNA. Sufficient reagents are provided to prepare uniquely barcoded DNA libraries from 384 research samples.
Note: The Ion Library TaqMan Quantitation Kit (Cat. No. 4468802) must be purchased separately.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is included in the Ion AmpliSeq Microbiome Health Research Kit, Library bundle?
The Ion AmpliSeq Microbiome Health Research Kit, Library bundle (Cat. No. A46495) consists of the Ion AmpliSeq Microbiome Health Research - 16S rRNA Gene Pool, the Ion AmpliSeq Microbiome Health Research - Target Species Pool, the Ion AmpliSeq Library Kit Plus, and the IonCode Barcode Adapters for the rapid preparation of barcoded sample libraries from bacterial DNA. Sufficient reagents are provided to prepare uniquely barcoded DNA libraries from 48 research samples.
Note: The Ion Library TaqMan Quantitation Kit (Cat. No. 4468802) must be purchased separately.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Can I use the AmpliSeq_Microbiome_Health_ Analysis plugin for Torrent Suite Software and Ion Reporter Software workflows for my custom 16S or target species AmpliSeq panel?
The Torrent Suite AmpliSeq_Microbiome_Health_ Analysis plugin and Ion Reporter Software workflows were developed and validated exclusively for the Ion AmpliSeq Microbiome Health Research Kit. Open source analysis tools may be available for your custom solutions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Can I customize the list of target species for my microbiome diversity research project?
Customization solutions are available through AmpliSeq custom designs (Ampliseq.com), where you can customize your own assays for any species of your interest. For details, please contact your local Field Applications Scientist (FAS).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Can the Ion AmpliSeq Microbiome Health Research Kit be used to investigate the human response to SARS-CoV-2 infection?
Several scientific publications have demonstrated the key role played by the gut microbial community in infection susceptibility and severity, including for SARS-CoV-2. The Ion AmpliSeq Microbiome Health Research Kit targets 73 species known to be associated with immune-related diseases, including a large number of known bacterial species with antiviral or immune-modulating properties.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.