Can the MS2 Phage Control within the TaqPath COVID-19 Combo Kit (Cat. No. A47814) be added in a different order to the reaction assay?
No, the MS2 Phage Control must be added last as described on page 24 in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019181_TaqPath_COVID-19_IFU_EUA.pdf).