Search
| Número de catálogo | Cantidad |
|---|---|
| A48397 | 500 tubes |
PrepFiler Capless Sample Tubes are intended for use with PrepFiler Lysep Columns for sample lysis and substrate removal in a single tube format. The sample tubes are capless to provide compatibility with the capped Lysep Columns. The innovative PrepFiler LySep Column dramatically streamlines the off-line lysis portion of the extraction method.
The PrepFiler Capless Sample Tube/Lysep Column combination is intended for use with the PrepFiler Automated Forensic DNA Extraction Kit, PrepFiler Forensic DNA Extraction Kit, PrepFiler BTA Automated Forensic DNA Extraction Kit, or PrepFiler BTA Forensic DNA Extraction Kit, which do not include plastic consumables, providing users the flexibility to order only the plastics consumables they need.
PrepFiler Forensic DNA Extraction kits use magnetic particles with an optimized multi-component surface chemistry to deliver superior DNA yield from most forensic sample types, including body fluids, stains and swabs of body fluids, bone, teeth, adhesive, and low-template samples.
The packaged tubes are treated with ethylene oxide.