Which deep-well plate would be a suitable alternative for the KingFisher 96 deep-well plate (Cat. No. 95040450)?
The following catalog numbers would be a suitable alternative: 95040460, A48305, 95040455 and A48424.
Search
View additional product information for KingFisher Plastic Consumables for SARS-CoV-2 Testing - FAQs (A48424)
1 product FAQs found