TrueMark™ Respiratory Panel 2.0, TaqMan™ Array card
Applied Biosystems™

TrueMark™ Respiratory Panel 2.0, TaqMan™ Array card

The TrueMark Respiratory Panel 2.0, TaqMan Array card, is an efficient, easy-to-use TaqMan Array card for the characterization of keyRead more
Have Questions?
Catalog NumberQuantity
A490471 Card
Catalog number A49047
Price (USD)
750.00
Each
Quantity:
1 Card
Price (USD)
750.00
Each
The TrueMark Respiratory Panel 2.0, TaqMan Array card, is an efficient, easy-to-use TaqMan Array card for the characterization of key respiratory tract microbes. The array card includes 44 TaqMan assays that have been optimized for detection of 41 respiratory tract viral, bacterial, and fungal microbes, including two assays that target SARS-CoV-2.

The array card also includes control assays for TaqMan Universal Extraction Control Organism (B. atrophaeus), TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control, the human RNase P RPPH1 gene, and the human 18S ribosomal RNA gene. For a complete list of assays included in the array card see the TrueMark Respiratory Panel 2.0, TaqMan Array card, product information sheet.

Features of this syndromic panel-based real-time PCR solution include:
• Extensive coverage—featuring the largest number of markers of pathogenic organisms of any test on the market
• Cost-effective workflow—lowest cost per sample when compared with other commercially available molecular solutions
• Multiple sample input types—including nasopharyngeal (NP) swabs, bronchoalveolar lavage (BAL), and nasopharyngeal aspirate
• Optimal throughput—run up to 8 samples per array card

Each TrueMark Respiratory Panel 2.0, TaqMan Array card, contains a select group of TaqMan microbial assays pre-configured in a 384-well micro-fluidic array card (48-assay format). Simply add the pre-amplified product to each port along with our TaqMan Fast Advanced Master Mix, no UNG, and nuclease-free water, centrifuge and seal the card, then load it into the instrument for results.

TaqMan assay designs and assay target sequences have undergone rigorous bioinformatics selection and analysis to maximize strain coverage and minimize potential for off‐target cross-reactivity. Qualified TaqMan assays also undergo performance testing to ensure that results are accurate and reproducible with high levels of analytical sensitivity and specificity.

The TrueMark Respiratory Panel 2.0, TaqMan Array card, is orderable as single cards and is intended for use with the QuantStudio 7 and QuantStudio 12K Flex systems with a TaqMan Array Card block.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
CompatibilityQuantStudio™ 7 and QuantStudio™ 12 K Flex systems with a TaqMan™ Array Card™ block
No. of ReactionsUp to 8 samples
Product LineTrueMark
Quantity1 Card
Shipping ConditionApproved for shipment at ambient temperature
Target41 respiratory plus 4 control
Target Organism ClassRespiratory
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C

Frequently asked questions (FAQs)

What is the shelf life of the TrueMark Respiratory Panel 2.0, TaqMan Array card (Cat. No. A49047)?

1 year from date of manufacture.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.