Product Image
Gibco™
Tube Clamps compatible with CTS™ Rotea™ Single-Use Kit
The Tube Clamps are compatible with the CTS Rotea Single-Use kits and are used to restrict fluid flow from connected bags.
Have Questions?
Número do catálogoQuantity
A49127100 Clamps
Número do catálogo A49127
Preço (BRL)
-
Quantity:
100 Clamps
The Tube Clamps are compatible with the CTS Rotea Single-Use kits and are used to restrict fluid flow from connected bags.
Especificações
Product LineCTS Rotea
Quantity100 Clamps
Product TypeTube Clamps
Unit SizeEach
Frequently asked questions (FAQs)
Do you have a calculation formula that allows us to estimate the max feed flow rate/processing time for the CTS Rotea System, depending on the cell diameter?
The Rotea Protocol Builder Software, which includes the Rotea Process Model and Simulator, is available for download from the Rotea website. Common cell types are already pre-loaded into the software, and custom cells can also be created. The CTS Rotea System can separate cells that have at least 4 µm difference in diameter. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs. Please visit our webpage (www.thermofisher.com/rotea) to see if the CTS Rotea System will work for your cell type, or submit a request for more information using our demo form (https://www.thermofisher.com/us/en/home/global/forms/life-science/rotea-counterflow-centrifugation-system-cell-therapy.html) to get connected with a Business Development Manager.
Are there plans for incorporating magnetic bead selection in the CTS Rotea System or other ways of automating magnetic bead selection?
Thermo Fisher Scientific strives to provide a catalog of tools to support our diverse customers' needs. If magnetic bead separation using the CTS Rotea System is of interest, Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
How does cell processing with Ficoll medium compare to using the CTS Rotea System?
Ficoll medium has many shortcomings and often does not scale well for cGMP manufacturing. Rotea PBMC isolation protocol takes less time and has been demonstrated to be capable of providing higher cell recovery compared to Ficoll medium. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
What is the best cryopreservative to use with the CTS Rotea System?
The CTS Rotea single-use kits are manufactured with DEHP-free PVC. Extractables and Leechables testing was conducted with 20% DMSO for up to 15 min. The CTS Rotea System has not been validated for fill-finish applications. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
Have you performed final formulation on the CTS Rotea System directly into cryopreservation media?
The CTS Rotea System has been demonstrated to be capable of washing cells from cryoprotectant into media and the reverse. The CTS Rotea System has not been validated for fill-finish applications. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs