96-Well Plate Retainer & Base Set, standard, for SeqStudio™ Flex, 4 sets - FAQs
96-Well Plate Retainer & Base Set, standard, for SeqStudio™ Flex, 4 sets - FAQs
View additional product information for 96-Well Plate Retainer & Base Set, for SeqStudio™ Flex - FAQs (A49316, A49495)
2 product FAQs found
When shouldn't I use the 3500 plate retainers on the SeqStudio Flex Series Genetic Analyzers?
If the automatic barcode workflow is enabled on the SeqStudio Flex Series Genetic Analyzer, using the 3500 plate retainers will cause a barcode error in the home screen.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
What is different about the SeqStudio Flex Series Genetic Analyzer plate retainers compared with the 3500 plate retainers?
The SeqStudio Flex Series Genetic Analyzer plate retainers have one opening on the side to allow barcode reading while the 3500 plate retainers have two openings on the side which can cover the plate barcode.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.