Product Image
Applied Biosystems™
96-Well Plate Retainer & Base Set, for SeqStudio™ Flex
This 96-well plate Retainer and Base Set is for use with 96-well sample plates on SeqStudio Flex Series genetic analyzers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate Compatibility
A49495Fast (0.1 mL) 96-well Sample Plates
A49316Standard 96-well plates
Catalog number A49495
Price (USD)
347.00
Each
Plate Compatibility:
Fast (0.1 mL) 96-well Sample Plates
Price (USD)
347.00
Each
This 96-well plate Retainer and Base Set is for use with 96-well sample plates on SeqStudio Flex Series genetic analyzers.
This 96-well plate Retainer and Base Set is for use with 96-well sample plates on SeqStudio Flex Series genetic analyzers.
Specifications
For Use With (Application)Sanger Sequencing and Fragment Analysis
For Use With (Equipment)SeqStudio™ Flex Series Genetic Analyzers
Plate CompatibilityFast (0.1 mL) 96-well Sample Plates
Quantity4 sets
Unit SizeEach
Frequently asked questions (FAQs)
When shouldn't I use the 3500 plate retainers on the SeqStudio Flex Series Genetic Analyzers?
If the automatic barcode workflow is enabled on the SeqStudio Flex Series Genetic Analyzer, using the 3500 plate retainers will cause a barcode error in the home screen.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
What is different about the SeqStudio Flex Series Genetic Analyzer plate retainers compared with the 3500 plate retainers?
The SeqStudio Flex Series Genetic Analyzer plate retainers have one opening on the side to allow barcode reading while the 3500 plate retainers have two openings on the side which can cover the plate barcode.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.