Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With (Application) | Mycoplasma detection |
Product Type | Purified genomic DNA |
Concentration | 100 GC/μL |
Sample Volume | 1 mL/species |
Shipping Condition | Dry Ice |
Species | Mycoplasma species: M. gallisepticum, M. synoviae, M. salivarium, M. hominis |
Product Line | MycoSEQ™ |
Unit Size | 1 kit |
For Use With (Application) | Mycoplasma detection |
Product Type | Purified genomic DNA |
Concentration | 100 GC/μL |
Sample Volume | 1 mL/species |
Shipping Condition | Dry Ice |
Species | Mycoplasma species: A. laidlawii, M. fermentans, M. hyorhinis, M. orale, M. arginini, M. pneumoniae |
Product Line | MycoSEQ™ |
Unit Size | 1 kit |
Catalog Number | Specifications | Unit Size | Species | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A49976 | 1 kit | Mycoplasma species: M. gallisepticum, M. synoviae, M. salivarium, M. hominis | Price: 6,180.00 Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A49973 | 1 kit | Mycoplasma species: A. laidlawii, M. fermentans, M. hyorhinis, M. orale, M. arginini, M. pneumoniae | Price: 9,270.00 Your Price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|