Detection Method: | Colorimetric |
---|---|
Label or Dye: | Digoxigenin, HRP (Horseradish Peroxidase) |
Substrate: | AEC |
Shipping Condition: | Wet Ice |
Substrate Type: | HRP (Horseradish Peroxidase) Substrate |
Target Molecule: | RNA |
Target Organism Class: | Epstein-Barr Virus (EBV) |
Target Specificity: | Target-Specific |
Product Line: | REMBRANDT® |