Detection Method: | Colorimetric |
---|---|
Label or Dye: | AP (Alkaline Phosphatase), Digoxigenin |
Substrate: | BCIP⁄NBT |
Shipping Condition: | Wet Ice |
Substrate Type: | AP (Alkaline Phosphatase) Substrate |
Target Molecule: | RNA |
Target Organism Class: | Epstein-Barr Virus (EBV) |
Target Specificity: | Target-Specific |
Product Line: | REMBRANDT® |