Product Image
Gibco™
Sterilized Luer Connectors compatible with CTS™ Rotea™ Single-Use Kit
The Sterilized Luer Connectors enable connection of bags to the Rotea Single-Use kit without the need for a tube welder. Each pack of connectors is double bagged and gamma irradiated.
Have Questions?
KatalognummerMenge
A5011010 x 32 Connectors
Katalognummer A50110
Preis (EUR)
624,00
Each
Menge:
10 x 32 Connectors
Preis (EUR)
624,00
Each
The Sterilized Luer Connectors, compatible with the CTS Rotea Single-Use kits, enable connection of bags without the need for a tube welder. Each pack of connectors is double bagged and gamma irradiated.
This product includes 10 packs of connectors. Each pack contains the following Luer connectors:
  • 8 x female Leur to barb
  • 8 x male Luer to barb
  • 8 x female Luer cap
  • 8 x male Luer plug
Specifications
Menge10 x 32 Connectors
ProduktlinieCTS Rotea
GekühltNo
Unit SizeEach
Häufig gestellte Fragen (FAQ)
Do you have a calculation formula that allows us to estimate the max feed flow rate/processing time for the CTS Rotea System, depending on the cell diameter?
The Rotea Protocol Builder Software, which includes the Rotea Process Model and Simulator, is available for download from the Rotea website. Common cell types are already pre-loaded into the software, and custom cells can also be created. The CTS Rotea System can separate cells that have at least 4 µm difference in diameter. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs. Please visit our webpage (www.thermofisher.com/rotea) to see if the CTS Rotea System will work for your cell type, or submit a request for more information using our demo form (https://www.thermofisher.com/us/en/home/global/forms/life-science/rotea-counterflow-centrifugation-system-cell-therapy.html) to get connected with a Business Development Manager.
Are there plans for incorporating magnetic bead selection in the CTS Rotea System or other ways of automating magnetic bead selection?
Thermo Fisher Scientific strives to provide a catalog of tools to support our diverse customers' needs. If magnetic bead separation using the CTS Rotea System is of interest, Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
How does cell processing with Ficoll medium compare to using the CTS Rotea System?
Ficoll medium has many shortcomings and often does not scale well for cGMP manufacturing. Rotea PBMC isolation protocol takes less time and has been demonstrated to be capable of providing higher cell recovery compared to Ficoll medium. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
What is the best cryopreservative to use with the CTS Rotea System?
The CTS Rotea single-use kits are manufactured with DEHP-free PVC. Extractables and Leechables testing was conducted with 20% DMSO for up to 15 min. The CTS Rotea System has not been validated for fill-finish applications. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
Have you performed final formulation on the CTS Rotea System directly into cryopreservation media?
The CTS Rotea System has been demonstrated to be capable of washing cells from cryoprotectant into media and the reverse. The CTS Rotea System has not been validated for fill-finish applications. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs