Medio CTS™ NK-Xpander™
Gibco™
Medio CTS™ NK-Xpander™
CTS NK-Xpander Medium is specifically formulated for superior expansion of human natural killer (hNK) cells for cell therapy applications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadFormato
A501900420 LBPC
A5019001500 mLFrasco
A50190025 LBPC
A50190055 LBPC, Bagged Supplement (100 mL)
A501900310 LBPC
Número de catálogo A5019004
Precio (EUR)
-
Cantidad:
20 L
Formato:
BPC
Gibco CTS NK-Xpander Medium is specifically formulated for superior expansion of human natural killer (hNK) cells for cell therapy applications. When hNK cells are enriched from human peripheral blood mononuclear cells (hPBMCs) and expanded in a feeder-free system using CTS NK-Xpander Medium supplemented with 5% hAB serum and 500U/mL IL-2, ≥90% of the hNK cells generated maintain CD56+/CD16+/CD3- surface marker expression. Furthermore, such hNK cells demonstrate degranulation and killing of K562 cancer cells when expanded in CTS NK-Xpander Medium.
Superior expansion potential
CTS NK-Xpander Medium is capable of producing a high yield of hNK cells without the need for feeder cells, providing robust expansion of enriched hNK cells from hPBMCs from qualified donors at day 14 to 15 versus other medium options. Cells maintain CD56+/CD16+/CD3- surface marker expression and demonstrate cytolytic capabilities when expanded in CTS NK-Xpander Medium.
Manufactured for cell therapy applications
CTS NK-Xpander Medium is manufactured without cytokines and growth factors to allow flexibility with regard to intended application. The medium does not include human or animal-derived components; however, supplementation with hAB serum is recommended for best results.
CTS quality
Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.* Documentation such as specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available to support your regulatory filing.
The products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified.
Closed-system compatibility
CTS NK-Xpander Medium is available in 500-mL-bottle and 5-L-bag formats for convenient process development and closed-system manufacturing scale-up. The 5-L bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable DEHP-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector providing versatile connection options for your manufacturing process, including compatibility with the CTS Rotea Counterflow Centrifugation System.
*Other aspects of USP <1043> will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP <1043> in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the AM).
Para su uso en investigación o fabricación de productos basados en células, genes o tejidos. PRECAUCIÓN: no diseñado para la administración directa en seres humanos o animales.
Especificaciones
Tipo de célulaNatural Killer (NK) cells
Para utilizar con (aplicación)Cell and Gene Therapy Research, Development, and Manufacturing
FormatoBPC
Calidad de fabricaciónISO 13485, MDSAP, FDA-registered, 21 CFR 820
Cantidad20 L
Duración de almacenamiento12 months from date of manufacture
Condiciones de envíoBasal: Ambient, Supplement: Dry Ice
ClasificaciónAnimal Origin
Culture TypeMammalian Cell Culture
FormularioLiquid
Serum LevelSerum-free
EsterilidadSterile-filtered
Sterilization MethodSterile-filtered
Sin aditivosNo Phenol Red
Unit SizeEach
Contenido y almacenamiento
• Basal Medium: Store at 2°C to 8°C and protect from light.
• Supplement: Store at -20°C and protect from light.
Preguntas frecuentes
Does CTS NK-Xpander Medium require activation beads for NK cell proliferation?
No. Unlike cytotoxic T cells, activation beads are not required for NK cell proliferation.
How can I address cell attachment in my NK cell culture?
Cell attachment is more frequently seen in flasks than plates. A cell scraper can be used to gently remove cells that lightly adhere to the flask surface. We recommend using non-tissue culture treated plates and flasks when expanding NK cells.
At what temperature should I store CTS NK-Xpander Basal Medium and CTS NK-Xpander Supplement (50X)?
We recommend storing CTS NK-Xpander Basal Medium at 2 to 8 degrees C, protected from light and CTS NK-Xpander Supplement (50X) at -20 to -5 degrees C, protected from light.
For how long can CTS NK-Xpander complete medium be used?
CTS NK-Xpander complete medium (CTS NK-Xpander Basal Medium, CTS NK-Xpander Supplement (50X), and 5% hAB serum) is stable for 3 weeks when stored in the dark at 2 to 8 degrees C.
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.