TrueMark™ Comprehensive Microbiota Control, 50 μL (5 x 107 copies/μL) - FAQs
TrueMark™ Comprehensive Microbiota Control, 50 μL (5 x 107 copies/μL) - FAQs
View additional product information for TrueMark™ Comprehensive Microbiota Control - FAQs (A50382, A50383)
2 product FAQs found
Which method/instrument was used to determine the concentration of the plasmid in the TrueMark Comprehensive Microbiota Control (Cat. No. A50382)?
The plasmid concentration was determined by means of digital PCR on the QuantStudio 3D Digital PCR System.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Are the targets in the TaqMan Comprehensive Microbiota Control (Cat. No. A50382) present as single copy?
Yes, they are. However, with the exception of Xeno and RNase P which are present as multiple copies.
Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.