resDNASEQ™ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kits
Applied Biosystems™

resDNASEQ™ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kits

The resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kit is a quantitative PCR (qPCR)-based system for the detection of residual kanamycin-resistant plasmids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
A50460Assay Kit and Sample Preparation
A50337Assay Kit only
Catalog number A50460
Price (USD)
4,395.00
Each
Includes:
Assay Kit and Sample Preparation
Price (USD)
4,395.00
Each
The resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kit is a quantitative PCR (qPCR)-based system for the detection of residual kanamycin-resistant plasmids used in the development of gene therapies, cell-based vaccines, and similar biotherapeutics. Reliable and rapid, the resDNASEQ system enables sensitive and specific quantitation of kanamycin-resistant plasmid DNA. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types, from in-process samples with different sample matrices to bulk drug substance.
Features of the resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kit include:
• Designed to quantitate residual plasmid DNA with any common alleles of kanamycin resistance gene
• Highly sensitive quantitation through innovative DNA-free master mix
• Highly sensitive quantitation using proven TaqMan real-time PCR technology
• Includes master mix, TaqMan primer/probe mix, kanamycin-resistant plasmid DNA standard, and sample preparation reagents

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

Integrated workflow solution
The resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kit with PrepSEQ Residual DNA Sample Preparation Kit is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing. The included PrepSEQ Residual DNA Sample Preparation Kit offers both a manual and automated sample preparation workflow. The resDNASEQ kit has been validated on the Applied Biosystems 7500 Fast Real-Time PCR System and the QuantStudio 5 Real-Time PCR System. Data analysis is streamlined using AccuSEQ Real-Time PCR Detection Software that provides accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance.

Easy to use
With the resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kit, you perform qPCR analysis to compare the amount of kanamycin-resistant plasmid DNA in your test samples to standard curves generated with known amounts of kanamycin-resistant plasmid DNA standard. The resDNASEQ kit includes the components you need to carry out these steps, from master mix to TaqMan primer/probe mix to DNA standard, as well as step-by-step instructions.

Specifications
FormatKit
IncludesAssay Kit and Sample Preparation
No. of Reactions100 Reactions
Product LineresDNASEQ
Quantity100 reactions
TargetKanamycin Resistant Plasmids
Detection MethodPrimer-probe
Label or DyeFAM
PCR MethodqPCR
Unit SizeEach
Contents & Storage
resDNASEQ KanR Control
• KanR DNA Control, 40 μL, 3.0 x 107 copies/μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ KanR DNA Real-Time PCR Reagents
• Environmental Master Mix v3 (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• KanR DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

PrepSEQ Nucleic Acid Extraction Kit
• Lysis Buffer (2 bottles, 50 mL/bottle)
• Binding Solution/Isopropanol (1 empty bottle)
• Wash Buffer Concentrate (2 bottles, 26 mL/bottle)
• Elution Buffer (1 bottle, 25 mL)
• Proteinase K Buffer (1 bottle, 50 mL)
• Magnetic Particles (2 tubes, 1.5 mL/tube)
• Proteinase K (20 mg/mL) (1 tube, 1.25 mL), store at or below –20°C
• PK Buffer II (1 bottle, 11 mL)
Store all at room temperature unless otherwise indicated.

PrepSEQ Residual DNA Sample Preparation Kit
• Proteinase K (20 mg/mL), 1 tube, 1.25 mL
• Yeast tRNA (10 mg/mL), 1 tube, 0.5 mL
• Glycogen (5 mg/mL), 2 tubes, 1.0 mL/tube
Store all at or below –20°C.