Pierce™ Microdialysis Plates
Thermo Scientific™

Pierce™ Microdialysis Plates

Simultaneously process 1 to 96 samples with 10 to 100 μL volumes with this convenient rapid dialysis system that is automation compatible and disposable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMWCOCapacity (Metric)Product Type
A5047010 kDa1 mLMicrodialysis Plate, 48-well
8826010 kDa0.1 mLMicrodialysis Plate, 96-well
88261Not Applicable2.2 mLDeep-well Plate, 96-well
882623.5 kDa0.1 mLMicrodialysis Plate, 96-well
A504622 kDa0.1 mLMicrodialysis Plate, 96-well
A504652 kDa0.3 mLMicrodialysis Plate, 96-well
A504662 kDa1 mLMicrodialysis Plate, 48-well
A504673.5 kDa0.3 mLMicrodialysis Plate, 96-well
A504683.5 kDa1 mLMicrodialysis Plate, 48-well
A5046910 kDa0.3 mLMicrodialysis Plate, 96-well
A5047120 kDa0.1 mLMicrodialysis Plate, 96-well
A5047220 kDa0.3 mLMicrodialysis Plate, 96-well
A5047320 kDa1 mLMicrodialysis Plate, 48-well
Catalog number A50470
Price (USD)
520.65
Online exclusive
560.00
Save 39.35
Each
MWCO:
10 kDa
Capacity (Metric):
1 mL
Product Type:
Microdialysis Plate, 48-well
Price (USD)
520.65
Online exclusive
560.00
Save 39.35
Each

Pierce 48-well and 96-well, 2K, 3.5K, 10K, and 20K MWCO microdialysis plates are an automation-compatible microplate with devices for the rapid dialysis of 1 to 96 samples with volumes of 10 to 1000 μL. Using the Pierce Microdialysis Plate, sample dialysis can be efficiently performed in a standard 96 deep-well plate with a minimal amount of buffer while still providing >95% removal of small molecules. The plate is compatible with standard 96-well laboratory equipment and automated liquid-handling systems, making it an ideal option for high-throughput applications.

Each sample processed with the Pierce Microdialysis Plate is dialyzed inside a single, removable dialysis device consisting of two regenerated cellulose membranes ∼2 mm apart. The short diffusion distance and large surface area facilitate rapid dialysis, with the removal of salts and small molecules often complete in 2–4 hours. Sample is recovered using standard laboratory micropipettes. The plate seals are made of high-quality polypropylene and help protect the samples from contamination and evaporation, especially during long-term storage.

Features of Pierce Microdialysis Plate:

  • High yield—greater than 90% protein recovery
  • Rapid—dialysis completed in 2–4 hours
  • Easy to use—sample loading and retrieval can be done with a standard pipette tip
  • Flexible—detachable 8-device strips, scalable from 1 to 96 samples
  • Automation-compatible—plate format conforms to the SBS Microplate Standard

Each 48-well and 96-well Pierce Microdialysis Plate comes pre-loaded with twelve 8-dialysis device strips. Each dialysis device strip can be removed from the plate and separated into individual microdialysis devices, allowing only the number of needed devices to be used and eliminating waste. The sample chamber inside each microdialysis device is formed by two low-binding regenerated cellulose membranes that are rated to retain proteins and other macromolecules >2,000 Daltons while allowing removal of buffer salts and contaminants.

Applications:

  • Protein and DNA desalting/buffer exchange
  • Removal of dyes after protein labeling
  • Enzyme activity/binding assays
  • Drug binding studies (also see Rapid Equilbrium Dialysis 'RED' devices)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)1 mL
MembraneCellulose Membrane, Dialysis
Product TypeMicrodialysis Plate, 48-well
Purification TargetBuffer Exchange
Quantity1 Plate Set
Sufficient ForDialysis of 1 to 48 samples, 150–1000 μL
FormatMicrodialysis Plate
MWCO10 kDa
Sample Volume (Metric)1 mL
Unit SizeEach
Contents & Storage
Product shipped at ambient temperature. Upon receipt, store at room temperature.

Frequently asked questions (FAQs)

Are cartridges from Microdialysis Plates sold separately?

We do not offer the cartridges from Microdialysis plates as stand-alone products. Each individual device from a Microdialysis Plate can be used in a microtube or plate, if the entire strip or plate is not needed.

What is the chemical compatibility of Microdialysis Plates?

Please refer to the Chemical Compatibility table in the respective product manual.

What are 48-well Microdialysis Plates?

The 48-well Microdialysis Plates are automation-compatible 48 deep-well microplates with microdialysis devices for the rapid dialysis of 1 to 48 samples. Each plate includes 8 detachable cartridges where each cartridge is a strip with 6 microdialysis devices.

Why did my sample volume increase during dialysis?

Most likely it‘s because osmotic pressure from dialyzing from high osmolarity to low osmolarity solutions causes a sample volume increase, as each sample component-including water-will move towards equilibrium concentration on both sides of the membrane.

What is the protein recovery after dialysis when using the 96-well Microdialysis plate?

Typical protein recovery is greater than 90% after dialysis.